missing translation for 'onlineSavingsMsg'
Learn More

2-Ethylhexyl bromide, 95%, Thermo Scientific Chemicals

55.88 € - 161.81 €

Chemical Identifiers

CAS 18908-66-2
Molecular Formula C8H17Br
Molecular Weight (g/mol) 193.13
MDL Number MFCD00000220
InChI Key NZWIYPLSXWYKLH-UHFFFAOYSA-N
Synonym 2-ethylhexyl bromide, 3-bromomethyl heptane, heptane, 3-bromomethyl, 1-bromo-2-ethylhexane, 1-bromo iso octane, 2-ethyl-bromohexane, 3-bromomethyl-heptane, 1-bromo-2 ethylhexane, 2-ethylhexylhydrobromide
PubChem CID 86804
IUPAC Name 3-(bromomethyl)heptane
SMILES CCCCC(CC)CBr
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10378880
View Documents Promotion Details
Thermo Scientific Acros
305331000
100 g Glass bottle
55.88 €
100g
Please to purchase this item. Need a web account? Register with us today!
10442261
View Documents Promotion Details
Thermo Scientific Acros
305335000
500 g Glass bottle
161.81 €
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

18908-66-2
193.13
NZWIYPLSXWYKLH-UHFFFAOYSA-N
86804
CCCCC(CC)CBr
C8H17Br
MFCD00000220
2-ethylhexyl bromide, 3-bromomethyl heptane, heptane, 3-bromomethyl, 1-bromo-2-ethylhexane, 1-bromo iso octane, 2-ethyl-bromohexane, 3-bromomethyl-heptane, 1-bromo-2 ethylhexane, 2-ethylhexylhydrobromide
3-(bromomethyl)heptane

Specifications

18908-66-2
100.0
1.0800g/mL
69°C
94% min. (GC)
CH3(CH2)3CH(C2H5)CH2Br
MFCD00000220
2-ethylhexyl bromide, 3-bromomethyl heptane, heptane, 3-bromomethyl, 1-bromo-2-ethylhexane, 1-bromo iso octane, 2-ethyl-bromohexane, 3-bromomethyl-heptane, 1-bromo-2 ethylhexane, 2-ethylhexylhydrobromide
NZWIYPLSXWYKLH-UHFFFAOYSA-N
3-(bromomethyl)heptane
86804
95%
2-Ethylhexyl bromide, 95%
94.0
Colorless to Yellow
75.0°C to 77.0°C (16.0 mmHg)
Authentic
C8H17Br
1.4530 to 1.4550
1.08
Solubility in water: insoluble.
CCCCC(CC)CBr
193.13
193.13
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

missing translation for 'einecsNumber' : 242-659-9

missing translation for 'tsca' : TSCA

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only