Learn More
2-Ethylanthraquinone, 98%, Thermo Scientific Chemicals
16.00 € - 59.90 €
Chemical Identifiers
CAS | 84-51-5 |
---|---|
Molecular Formula | C16H12O2 |
Molecular Weight (g/mol) | 236.27 |
MDL Number | MFCD00001237 |
InChI Key | SJEBAWHUJDUKQK-UHFFFAOYSA-N |
Synonym | 2-ethylanthraquinone, 2-ethyl anthraquinone, 2-ethyl-9,10-anthraquinone, anthraquinone, 2-ethyl, 9,10-anthracenedione, 2-ethyl, usaf so-1, beta-ethylanthraquinone, unii-59yj81qzkd, 59yj81qzkd, 2-ethylanthra-9,10-quinone |
PubChem CID | 6772 |
IUPAC Name | 2-ethylanthracene-9,10-dione |
SMILES | CCC1=CC2=C(C=C1)C(=O)C3=CC=CC=C3C2=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11317186
|
Thermo Scientific Alfa Aesar
L07206.06 |
5 g |
22.31 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11327186
|
Thermo Scientific Alfa Aesar
L07206.22 |
100 g | N/A | ||||||
11337186
|
Thermo Scientific Alfa Aesar
L07206.36 |
500 g | N/A | ||||||
Description
2-Ethylanthraquinone can be used as a photoinitiator of crosslinking or degradation of polyethylene. It is used to produce hydrogen peroxide by hydrogenation with the help of Pd/polyaniline as the catalysts.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Ethylanthraquinone can be used as a photoinitiator of crosslinking or degradation of polyethylene. It is used to produce hydrogen peroxide by hydrogenation with the help of Pd/polyaniline as the catalysts.
Solubility
Insoluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Chemical Identifiers
84-51-5 | |
236.27 | |
SJEBAWHUJDUKQK-UHFFFAOYSA-N | |
6772 | |
CCC1=CC2=C(C=C1)C(=O)C3=CC=CC=C3C2=O |
C16H12O2 | |
MFCD00001237 | |
2-ethylanthraquinone, 2-ethyl anthraquinone, 2-ethyl-9,10-anthraquinone, anthraquinone, 2-ethyl, 9,10-anthracenedione, 2-ethyl, usaf so-1, beta-ethylanthraquinone, unii-59yj81qzkd, 59yj81qzkd, 2-ethylanthra-9,10-quinone | |
2-ethylanthracene-9,10-dione |
Specifications
84-51-5 | |
180°C to 190°C | |
C16H12O2 | |
1969873 | |
Insoluble in water. | |
CCC1=CC2=C(C=C1)C(=O)C3=CC=CC=C3C2=O | |
236.27 | |
236.27 | |
2-Ethylanthraquinone |
107°C to 111°C | |
>210°C (410°F) | |
MFCD00001237 | |
2-ethylanthraquinone, 2-ethyl anthraquinone, 2-ethyl-9,10-anthraquinone, anthraquinone, 2-ethyl, 9,10-anthracenedione, 2-ethyl, usaf so-1, beta-ethylanthraquinone, unii-59yj81qzkd, 59yj81qzkd, 2-ethylanthra-9,10-quinone | |
SJEBAWHUJDUKQK-UHFFFAOYSA-N | |
2-ethylanthracene-9,10-dione | |
6772 | |
98% |
Safety and Handling
missing translation for 'einecsNumber' : 201-535-4
missing translation for 'rtecsNumber' : CB0525000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.