missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(Dimethylaminomethyl)-1-cyclohexanone hydrochloride, 98%, Thermo Scientific Chemicals
60.30 € - 184.00 €
Chemical Identifiers
CAS | 42036-65-7 |
---|---|
Molecular Formula | C9H17NO·HCl |
Molecular Weight (g/mol) | 191.7 |
MDL Number | MFCD00102580 |
InChI Key | CLVHTSWMNNSUSH-UHFFFAOYSA-N |
Synonym | 2-dimethylamino methyl cyclohexanone hydrochloride, 2-dimethylaminomethyl-1-cyclohexanone hydrochloride, dimethyl 2-oxocyclohexyl methyl ammonium chloride, 2-dimethylamino methyl cyclohexan-1-one hydrochloride, 2-dimethylaminomethyl-cyclohexanone, 2-dimethylamino methyl cyclohexan-1-one, chloride, tramadol related compound b, 2-dimethylaminomethyl-cyclohexanone hydrochloride, tramadol related compound b usp |
PubChem CID | 359482 |
IUPAC Name | 2-[(dimethylamino)methyl]cyclohexan-1-one;hydrochloride |
SMILES | CN(C)CC1CCCCC1=O.Cl |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10737972
|
Thermo Scientific Acros
335620010 |
1 g | Glass bottle |
60.30 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10449412
|
Thermo Scientific Acros
335620050 |
5 g | Glass bottle |
184.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
42036-65-7 | |
191.7 | |
CLVHTSWMNNSUSH-UHFFFAOYSA-N | |
359482 | |
CN(C)CC1CCCCC1=O.Cl |
C9H17NO·HCl | |
MFCD00102580 | |
2-dimethylamino methyl cyclohexanone hydrochloride, 2-dimethylaminomethyl-1-cyclohexanone hydrochloride, dimethyl 2-oxocyclohexyl methyl ammonium chloride, 2-dimethylamino methyl cyclohexan-1-one hydrochloride, 2-dimethylaminomethyl-cyclohexanone, 2-dimethylamino methyl cyclohexan-1-one, chloride, tramadol related compound b, 2-dimethylaminomethyl-cyclohexanone hydrochloride, tramadol related compound b usp | |
2-[(dimethylamino)methyl]cyclohexan-1-one;hydrochloride |
Specifications
42036-65-7 | |
100.0 | |
White | |
98% | |
MFCD00102580 | |
Solubility in water: soluble in water. Other solubilities: soluble in methanol,ethanol | |
CN(C)CC1CCCCC1=O.Cl | |
191.7 | |
191.7 | |
Crystalline Powder |
97.5 | |
147°C to 151°C | |
Authentic | |
C9H17NO·HCl | |
2-dimethylamino methyl cyclohexanone hydrochloride, 2-dimethylaminomethyl-1-cyclohexanone hydrochloride, dimethyl 2-oxocyclohexyl methyl ammonium chloride, 2-dimethylamino methyl cyclohexan-1-one hydrochloride, 2-dimethylaminomethyl-cyclohexanone, 2-dimethylamino methyl cyclohexan-1-one, chloride, tramadol related compound b, 2-dimethylaminomethyl-cyclohexanone hydrochloride, tramadol related compound b usp | |
CLVHTSWMNNSUSH-UHFFFAOYSA-N | |
2-[(dimethylamino)methyl]cyclohexan-1-one;hydrochloride | |
359482 | |
98% | |
2-(Dimethylaminomethyl)-1-cyclohexanone hydrochloride, 98% |
Safety and Handling
missing translation for 'tsca' : TSCA
Product Identifier
- 2-(Dimethylaminomethyl)-1-cyclohexanone hydrochloride
RUO – Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title