Learn More
2-Dimethylaminoethyl chloride hydrochloride, 99%, Thermo Scientific Chemicals
Chemical Identifiers
CAS | 4584-46-7, 107-99-3 |
---|---|
Molecular Formula | C4H10ClN·HCl |
Molecular Weight (g/mol) | 144.05 |
MDL Number | MFCD00012516 |
InChI Key | LQLJZSJKRYTKTP-UHFFFAOYSA-N |
Synonym | 2-dimethylaminoethyl chloride hydrochloride, 2-chloro-n,n-dimethylethanamine hydrochloride, 2-dimethylamino ethyl chloride hydrochloride, 2-chloroethyl dimethylamine hydrochloride, 2-chloro-n,n-dimethylethylamine hydrochloride, dimethylaminoethyl chloride hydrochloride, 2-chloroethyldimethylamine hydrochloride, 1-chloro-2-dimethylaminoethane hydrochloride, 2-dimethylamino ethylchloride hydrochloride, n-2-chloroethyl dimethylamine hydrochloride |
PubChem CID | 11636816 |
ChEBI | CHEBI:78153 |
IUPAC Name | 2-chloro-N,N-dimethylethanamine;hydrochloride |
SMILES | CN(C)CCCl.Cl |
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Alkylation reagentChemical Identifiers
4584-46-7, 107-99-3 | |
144.05 | |
LQLJZSJKRYTKTP-UHFFFAOYSA-N | |
11636816 | |
2-chloro-N,N-dimethylethanamine;hydrochloride |
C4H10ClN·HCl | |
MFCD00012516 | |
2-dimethylaminoethyl chloride hydrochloride, 2-chloro-n,n-dimethylethanamine hydrochloride, 2-dimethylamino ethyl chloride hydrochloride, 2-chloroethyl dimethylamine hydrochloride, 2-chloro-n,n-dimethylethylamine hydrochloride, dimethylaminoethyl chloride hydrochloride, 2-chloroethyldimethylamine hydrochloride, 1-chloro-2-dimethylaminoethane hydrochloride, 2-dimethylamino ethylchloride hydrochloride, n-2-chloroethyl dimethylamine hydrochloride | |
CHEBI:78153 | |
CN(C)CCCl.Cl |
Specifications
4584-46-7, 107-99-3 | |
100.0 | |
Brown-Yellow to White | |
99% | |
(CH3)2NCH2CH2Cl·HCl | |
04, 133 | |
Solubility in water: 2000g/L (20°C). Other solubilities: soluble in alcohol (20°C) | |
CN(C)CCCl.Cl | |
144.05 | |
CHEBI:78153 | |
99% | |
2-Dimethylaminoethyl chloride hydrochloride, 99% |
98.5 | |
203.0°C to 210.0°C | |
Authentic | |
C4H10ClN·HCl | |
MFCD00012516 | |
2-dimethylaminoethyl chloride hydrochloride, 2-chloro-n,n-dimethylethanamine hydrochloride, 2-dimethylamino ethyl chloride hydrochloride, 2-chloroethyl dimethylamine hydrochloride, 2-chloro-n,n-dimethylethylamine hydrochloride, dimethylaminoethyl chloride hydrochloride, 2-chloroethyldimethylamine hydrochloride, 1-chloro-2-dimethylaminoethane hydrochloride, 2-dimethylamino ethylchloride hydrochloride, n-2-chloroethyl dimethylamine hydrochloride | |
LQLJZSJKRYTKTP-UHFFFAOYSA-N | |
2-chloro-N,N-dimethylethanamine;hydrochloride | |
11636816 | |
144.05 | |
Crystalline Powder or Crystals |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Suspected of causing genetic defects.
May cause damage to organs through prolonged or repeated exposure.
Toxic to aquatic life with long lasting effects.
GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
Immediately call a
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 224-970-1
missing translation for 'rtecsNumber' : KQ9020000
missing translation for 'tsca' : TSCA
- 2-Dimethylaminoethyl chloride hydrochloride
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 2
- Germ cell mutagenicity Category 2
- Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
- H341-Suspected of causing genetic defects.
- H373-May cause damage to organs.
- H411-Toxic to aquatic life with long lasting effects.
- H302-Harmful if swallowed.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.