missing translation for 'onlineSavingsMsg'
Learn More

2-Diisopropylaminoethyl chloride hydrochloride, 98%, Thermo Scientific Chemicals

Chemical Identifiers

CAS 4261-68-1
Molecular Formula C8H19Cl2N
Molecular Weight (g/mol) 200.15
MDL Number MFCD00012496
InChI Key IUSXYVRFJVAVOB-UHFFFAOYSA-N
Synonym 2-diisopropylaminoethyl chloride hydrochloride, 2-diisopropylamino ethyl chloride hydrochloride, unii-6rmr38fceq, 2-chloroethyldiisopropylamine hydrochloride, 6rmr38fceq, 2-propanamine, n-2-chloroethyl-n-1-methylethyl-, hydrochloride, 2-chloroethyldiisopropylammonium chloride, 2-chloroethyl diisopropylamine hydrochloride, n-chloroethyl diisopropylamine hydrochloride, beta-chloroethyl diisopropylamine hydrochloride
PubChem CID 77942
SMILES [H+].[Cl-].CC(C)N(CCCl)C(C)C
View More Specs

Description

Description

2-Diisopropylaminoethyl chloride hydrochloride is used as an intermediate in pharmaceuticals, agrochemicals, dyestuffs and in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Diisopropylaminoethyl chloride hydrochloride is used as an intermediate in pharmaceuticals, agrochemicals, dyestuffs and in organic synthesis.

Solubility
Soluble in water.

Notes
Hygroscopic. Air sensitive. Incompatible with bases and oxidizing agents.
Specifications

Chemical Identifiers

4261-68-1
200.15
IUSXYVRFJVAVOB-UHFFFAOYSA-N
77942
C8H19Cl2N
MFCD00012496
2-diisopropylaminoethyl chloride hydrochloride, 2-diisopropylamino ethyl chloride hydrochloride, unii-6rmr38fceq, 2-chloroethyldiisopropylamine hydrochloride, 6rmr38fceq, 2-propanamine, n-2-chloroethyl-n-1-methylethyl-, hydrochloride, 2-chloroethyldiisopropylammonium chloride, 2-chloroethyl diisopropylamine hydrochloride, n-chloroethyl diisopropylamine hydrochloride, beta-chloroethyl diisopropylamine hydrochloride
[H+].[Cl-].CC(C)N(CCCl)C(C)C

Specifications

4261-68-1
C8H19Cl2N
UN2811
Hygroscopic
Soluble in water.
[H+].[Cl-].CC(C)N(CCCl)C(C)C
200.15
200.15
2-Diisopropylaminoethyl chloride hydrochloride
131°C to 134°C
MFCD00012496
3551603
2-diisopropylaminoethyl chloride hydrochloride, 2-diisopropylamino ethyl chloride hydrochloride, unii-6rmr38fceq, 2-chloroethyldiisopropylamine hydrochloride, 6rmr38fceq, 2-propanamine, n-2-chloroethyl-n-1-methylethyl-, hydrochloride, 2-chloroethyldiisopropylammonium chloride, 2-chloroethyl diisopropylamine hydrochloride, n-chloroethyl diisopropylamine hydrochloride, beta-chloroethyl diisopropylamine hydrochloride
IUSXYVRFJVAVOB-UHFFFAOYSA-N
N-(2-chloroethyl)-N-propan-2-ylpropan-2-amine;hydrochloride
77942
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H300-H310-H330-H318-H315-H335
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Causes serious eye damage.
Causes skin irritation.
May cause respiratory irritation.

P260-P264b-P270-P271-P280-P284-P301+P310-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P361-P363-P501c

H301+H311-H315-H319-H330-H335

missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

missing translation for 'einecsNumber' : 224-238-1

missing translation for 'rtecsNumber' : TX1521900

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Certificates

RUO – Research Use Only