missing translation for 'onlineSavingsMsg'
Learn More

2-Diethylaminoethylchloride hydrochloride, 99.5%, Thermo Scientific Chemicals

Chemical Identifiers

CAS 869-24-9
Molecular Formula C6H15Cl2N
Molecular Weight (g/mol) 172.09
MDL Number MFCD00012519
InChI Key RAGSWDIQBBZLLL-UHFFFAOYSA-N
Synonym 2-chloro-n,n-diethylethanamine hydrochloride, 2-diethylaminoethylchloride hydrochloride, 2-chlorotriethylamine hydrochloride, 2-chloro-n,n-diethylethylamine hydrochloride, 2-diethylamino ethyl chloride hydrochloride, diethylaminoethyl chloride hydrochloride, n,n-diethyl-2-chloroethylamine hydrochloride, 2-chloroethyl diethylamine hydrochloride, unii-ycy6eqt3wa, ethanamine, 2-chloro-n,n-diethyl-, hydrochloride
PubChem CID 13363
IUPAC Name 2-chloro-N,N-diethylethanamine;hydrochloride
SMILES [H+].[Cl-].CCN(CC)CCCl
View More Specs

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

869-24-9
172.09
RAGSWDIQBBZLLL-UHFFFAOYSA-N
13363
[H+].[Cl-].CCN(CC)CCCl
C6H15Cl2N
MFCD00012519
2-chloro-n,n-diethylethanamine hydrochloride, 2-diethylaminoethylchloride hydrochloride, 2-chlorotriethylamine hydrochloride, 2-chloro-n,n-diethylethylamine hydrochloride, 2-diethylamino ethyl chloride hydrochloride, diethylaminoethyl chloride hydrochloride, n,n-diethyl-2-chloroethylamine hydrochloride, 2-chloroethyl diethylamine hydrochloride, unii-ycy6eqt3wa, ethanamine, 2-chloro-n,n-diethyl-, hydrochloride
2-chloro-N,N-diethylethanamine;hydrochloride

Specifications

869-24-9
100.0
White to Beige
Authentic
C6H15Cl2N
MFCD00012519
2-chloro-n,n-diethylethanamine hydrochloride, 2-diethylaminoethylchloride hydrochloride, 2-chlorotriethylamine hydrochloride, 2-chloro-n,n-diethylethylamine hydrochloride, 2-diethylamino ethyl chloride hydrochloride, diethylaminoethyl chloride hydrochloride, n,n-diethyl-2-chloroethylamine hydrochloride, 2-chloroethyl diethylamine hydrochloride, unii-ycy6eqt3wa, ethanamine, 2-chloro-n,n-diethyl-, hydrochloride
RAGSWDIQBBZLLL-UHFFFAOYSA-N
2-chloro-N,N-diethylethanamine;hydrochloride
13363
99.5%
2-Diethylaminoethylchloride hydrochloride
99.4
208.0°C to 212.0°C
1% max. (105°C, 3 hrs)
99.5%
(C2H5)2NCH2CH2Cl·HCl
04, III, 240
Solubility in water: 2000g/L (20°C)
[H+].[Cl-].CCN(CC)CCCl
172.09
172.1
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Causes severe skin burns and eye damage.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 212-786-4

Product Identifier
  • 2-Diethylaminoethylchloride hydrochloride

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 C

Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H310-Fatal in contact with skin.
  • H330-Fatal if inhaled.
  • H300-Fatal if swallowed.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P330-Rinse mouth.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only