Learn More
Thermo Scientific Chemicals 2'-Deoxyadenosine, 99%
38.10 € - 466.00 €
Chemical Identifiers
CAS | 958-09-8 |
---|---|
Molecular Formula | C10H13N5O3 |
Molecular Weight (g/mol) | 251.25 |
MDL Number | MFCD00005754 |
InChI Key | OLXZPDWKRNYJJZ-RRKCRQDMSA-N |
Synonym | 2'-deoxyadenosine, deoxyadenosine, 2-deoxyadenosine, 2r,3s,5r-5-6-amino-9h-purin-9-yl-2-hydroxymethyl tetrahydrofuran-3-ol, adenyldeoxyriboside, adenine deoxyribonucleoside, adenine deoxyribose, desoxyadenosine, ccris 1782, unii-p582c98ulc |
PubChem CID | 13730 |
ChEBI | CHEBI:17256 |
IUPAC Name | (2R,3S,5R)-5-(6-aminopurin-9-yl)-2-(hydroxymethyl)oxolan-3-ol |
SMILES | NC1=C2N=CN([C@H]3C[C@H](O)[C@@H](CO)O3)C2=NC=N1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15486039
|
Thermo Scientific Chemicals
J63886.06 |
5 g |
38.10 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15496039
|
Thermo Scientific Chemicals
J63886.14 |
25 g |
170.00 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15406049
|
Thermo Scientific Chemicals
J63886.18 |
50 g |
258.00 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15416049
|
Thermo Scientific Chemicals
J63886.22 |
100 g |
466.00 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
2'-Deoxyadenosine is used in paradoxical stimulation study of human sperm motility. It plays an essential role as sperm motility stimulants used in sperm micro-injection on mouse and human embryo development. It finds an application in some cells as an energy source under energy stress conditions. It is involved in various biological processes to compare the functions of adenosine analogues.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2′-Deoxyadenosine is used in paradoxical stimulation study of human sperm motility. It plays an essential role as sperm motility stimulants used in sperm micro-injection on mouse and human embryo development. It finds an application in some cells as an energy source under energy stress conditions. It is involved in various biological processes to compare the functions of adenosine analogues.
Solubility
Soluble in water and caustic soda.
Notes
Air sensitive. Keep container tightly closed in a dry and well-ventilated place.
Chemical Identifiers
958-09-8 | |
251.25 | |
OLXZPDWKRNYJJZ-RRKCRQDMSA-N | |
13730 | |
(2R,3S,5R)-5-(6-aminopurin-9-yl)-2-(hydroxymethyl)oxolan-3-ol |
C10H13N5O3 | |
MFCD00005754 | |
2'-deoxyadenosine, deoxyadenosine, 2-deoxyadenosine, 2r,3s,5r-5-6-amino-9h-purin-9-yl-2-hydroxymethyl tetrahydrofuran-3-ol, adenyldeoxyriboside, adenine deoxyribonucleoside, adenine deoxyribose, desoxyadenosine, ccris 1782, unii-p582c98ulc | |
CHEBI:17256 | |
NC1=C2N=CN([C@H]3C[C@H](O)[C@@H](CO)O3)C2=NC=N1 |
Specifications
958-09-8 | |
C10H13N5O3 | |
2'-deoxyadenosine, deoxyadenosine, 2-deoxyadenosine, 2r,3s,5r-5-6-amino-9h-purin-9-yl-2-hydroxymethyl tetrahydrofuran-3-ol, adenyldeoxyriboside, adenine deoxyribonucleoside, adenine deoxyribose, desoxyadenosine, ccris 1782, unii-p582c98ulc | |
OLXZPDWKRNYJJZ-RRKCRQDMSA-N | |
NC1=C2N=CN([C@H]3C[C@H](O)[C@@H](CO)O3)C2=NC=N1 | |
251.25 | |
CHEBI:17256 | |
99% | |
2'-Deoxyadenosine |
White | |
MFCD00005754 | |
Soluble in water and caustic soda. | |
−25° (c=0.5 in Water) | |
(2R,3S,5R)-5-(6-aminopurin-9-yl)-2-(hydroxymethyl)oxolan-3-ol | |
13730 | |
251.24 | |
Powder |
Safety and Handling
P264b-P270-P301+P312-P330-P501c
H302
missing translation for 'einecsNumber' : 213-488-7
missing translation for 'rtecsNumber' : AU7358650
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only