missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals 2-Deoxy-D-ribose, 99%
78.60 € - 259.00 €
Chemical Identifiers
CAS | 533-67-5 |
---|---|
Molecular Formula | C5H10O4 |
Molecular Weight (g/mol) | 134.13 |
MDL Number | MFCD00135904 |
InChI Key | PDWIQYODPROSQH-GLULIZFNNA-N |
Synonym | 2-deoxy-d-ribose, thyminose, 3s,4r-3,4,5-trihydroxypentanal, deoxyribose, 2-deoxy-d-erythro-pentose, 2-deoxyribose, d-drib, 2-deoxy-d-arabinose, unii-lsw4h01241, 2-deoxy-d-erythropentose |
PubChem CID | 5460005 |
ChEBI | CHEBI:28816 |
IUPAC Name | (3S,4R)-3,4,5-trihydroxypentanal |
SMILES | OC[C@H]1OC(O)C[C@@H]1O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10245720
|
Thermo Scientific Chemicals
153280050 |
5 g | Glass bottle |
78.60 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10746001
|
Thermo Scientific Chemicals
153280250 |
25 g | Glass bottle |
259.00 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
533-67-5 | |
134.13 | |
PDWIQYODPROSQH-GLULIZFNNA-N | |
5460005 | |
(3S,4R)-3,4,5-trihydroxypentanal |
C5H10O4 | |
MFCD00135904 | |
2-deoxy-d-ribose, thyminose, 3s,4r-3,4,5-trihydroxypentanal, deoxyribose, 2-deoxy-d-erythro-pentose, 2-deoxyribose, d-drib, 2-deoxy-d-arabinose, unii-lsw4h01241, 2-deoxy-d-erythropentose | |
CHEBI:28816 | |
OC[C@H]1OC(O)C[C@@H]1O |
Specifications
533-67-5 | |
100.0 | |
White to Yellow | |
1.0% max. (40°C, 2 hrs) (vacuum) | |
98.5% min | |
MFCD00135904 | |
-57 | |
Solubility in water: soluble. Other solubilities: soluble in pyridine,slightly soluble in alcohol | |
OC[C@H]1OC(O)C[C@@H]1O | |
134.13 | |
CHEBI:28816 | |
99% | |
2-Deoxy-D-ribose, 99% |
98.5 | |
91°C | |
0.5% max. | |
Authentic | |
C5H10O4 | |
15, 2909 | |
2-deoxy-d-ribose, thyminose, 3s,4r-3,4,5-trihydroxypentanal, deoxyribose, 2-deoxy-d-erythro-pentose, 2-deoxyribose, d-drib, 2-deoxy-d-arabinose, unii-lsw4h01241, 2-deoxy-d-erythropentose | |
PDWIQYODPROSQH-GLULIZFNNA-N | |
(3S,4R)-3,4,5-trihydroxypentanal | |
5460005 | |
134.13 | |
Crystalline Powder |
Safety and Handling
missing translation for 'einecsNumber' : 208-573-0
missing translation for 'rtecsNumber' : SB7230000
missing translation for 'tsca' : TSCA
RUO – Research Use Only