missing translation for 'onlineSavingsMsg'
Learn More

2-Cyclohexyloxypyridine-5-carboxaldehyde, 97%, Thermo Scientific™

Product Code. 11434590
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 916792-16-0
Molecular Formula C12H15NO2
Molecular Weight (g/mol) 205.257
MDL Number MFCD08060944
InChI Key KTDSNAKFASLMHF-UHFFFAOYSA-N
Synonym 2-cyclohexyloxypyridine-5-carboxaldehyde, 6-cyclohexyloxy nicotinaldehyde, 6-cyclohexyloxy pyridine-3-carbaldehyde, 6-cyclohexyl-oxy pyridine-3-carbaldehyde
PubChem CID 42553152
IUPAC Name 6-cyclohexyloxypyridine-3-carbaldehyde
SMILES C1CCC(CC1)OC2=NC=C(C=C2)C=O

Specifications

Quantity 1 g
Sensitivity Air and light sensitive
Formula Weight 205.26
Percent Purity 97%
Chemical Name or Material 2-Cyclohexyloxypyridine-5-carboxaldehyde
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'