Learn More
2-Cyclohexylethylamine, 97%, Thermo Scientific Chemicals
114.57 € - 393.96 €
Chemical Identifiers
CAS | 4442-85-7 |
---|---|
Molecular Formula | C8H17N |
Molecular Weight (g/mol) | 127.23 |
MDL Number | MFCD00058668 |
InChI Key | HFACYWDPMNWMIW-UHFFFAOYSA-N |
Synonym | 2-cyclohexylethylamine, 2-cyclohexyl-ethylamine, cyclohexaneethanamine, cyclohexaneethylamine, 2-cyclohexylethan-1-amine, cyclohexaneethanamine 9ci, 2-aminoethylcyclohexane, ethylamine, 2-cyclohexyl, chembl19428, 2-cyclohexylethyl amine hydrochloride |
PubChem CID | 20509 |
IUPAC Name | 2-cyclohexylethanamine |
SMILES | NCCC1CCCCC1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11397549
|
Thermo Scientific Alfa Aesar
H27987.03 |
1 g |
114.57 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11307559
|
Thermo Scientific Alfa Aesar
H27987.06 |
5 g |
393.96 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
In the resolution system of (RS)-1-cyclohexylethylamine (CHEA) with (S)-mandelic acid (MA), the DCR (dielectrically controlled resolution) phenomenon was clearly observed.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIn the resolution system of (RS)-1-cyclohexylethylamine (CHEA) with (S)-mandelic acid (MA), the DCR (dielectrically controlled resolution) phenomenon was clearly observed.
Notes
Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air. Protect from direct light.
Chemical Identifiers
4442-85-7 | |
127.23 | |
HFACYWDPMNWMIW-UHFFFAOYSA-N | |
20509 | |
NCCC1CCCCC1 |
C8H17N | |
MFCD00058668 | |
2-cyclohexylethylamine, 2-cyclohexyl-ethylamine, cyclohexaneethanamine, cyclohexaneethylamine, 2-cyclohexylethan-1-amine, cyclohexaneethanamine 9ci, 2-aminoethylcyclohexane, ethylamine, 2-cyclohexyl, chembl19428, 2-cyclohexylethyl amine hydrochloride | |
2-cyclohexylethanamine |
Specifications
4442-85-7 | |
MFCD00058668 | |
Air sensitive | |
HFACYWDPMNWMIW-UHFFFAOYSA-N | |
2-cyclohexylethanamine | |
20509 | |
97% |
C8H17N | |
UN2735 | |
2-cyclohexylethylamine, 2-cyclohexyl-ethylamine, cyclohexaneethanamine, cyclohexaneethylamine, 2-cyclohexylethan-1-amine, cyclohexaneethanamine 9ci, 2-aminoethylcyclohexane, ethylamine, 2-cyclohexyl, chembl19428, 2-cyclohexylethyl amine hydrochloride | |
NCCC1CCCCC1 | |
127.23 | |
127.23 | |
2-Cyclohexylethylamine |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
missing translation for 'einecsNumber' : 224-673-7
missing translation for 'rtecsNumber' : KR3325000
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only