missing translation for 'onlineSavingsMsg'
Learn More

2-Cyclohexylethyl bromide, 98%, Thermo Scientific Chemicals

Product Code. 10570384
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1647-26-3
Molecular Formula C8H15Br
Molecular Weight (g/mol) 191.11
MDL Number MFCD00019398
InChI Key JRQAAYVLPPGEHT-UHFFFAOYSA-N
Synonym 2-bromoethyl cyclohexane, 2-cyclohexylethyl bromide, cyclohexane, 2-bromoethyl, 2-cyclohexylethylbromide, 1-bromo-2-cyclohexylethane, cyclohexylethylbromide, bromoethyl cyclohexane, cyclohexylethyl bromide, acmc-1bzct, 1-bromo-2cyclohexylethane
PubChem CID 15440
IUPAC Name 2-bromoethylcyclohexane
SMILES BrCCC1CCCCC1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point -57°C
Color Colorless to Yellow
Density 1.2210g/mL
Boiling Point 209°C
Flash Point 96°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula C6H11(CH2)2Br
Refractive Index 1.4890 to 1.4910 (20°C, 589nm)
Quantity 1 g
Specific Gravity 1.221
Formula Weight 191.11
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-Cyclohexylethyl bromide
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.