missing translation for 'onlineSavingsMsg'
Learn More

2-Cyclohexen-1-one, 97%, Thermo Scientific Chemicals

Product Code. 11492777
Click to view available options
Quantity:
25 g
100 g
500 g
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

2-Cyclohexen-1-one is used as intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Cyclohexen-1-one is used as intermediates.

Solubility
Soluble in water, alcohols, benzene and acetone.

Notes
Store away from oxidizing agents and strong bases. Incompatible with reducing agents and bases.
TRUSTED_SUSTAINABILITY

Identifiants chimiques

CAS 930-68-7
Molecular Formula C6H8O
Molecular Weight (g/mol) 96.13
MDL Number MFCD00001577
InChI Key FWFSEYBSWVRWGL-UHFFFAOYSA-N
Synonym 2-cyclohexen-1-one, cyclohex-2-enone, 2-cyclohexenone, cyclohexenone, 3-oxocyclohexene, 1-cyclohexen-3-one, cyclohexen-3-one, cyclohexen-1-one, 2-cyclohexenone-1, 2-cyclohexene-1-one
PubChem CID 13594
ChEBI CHEBI:15977
IUPAC Name cyclohex-2-en-1-one
SMILES O=C1CCCC=C1

Spécification

Melting Point -53°C
Density 0.993
Boiling Point 167°C to 169°C
Flash Point 56°C (132°F)
Refractive Index 1.488
Quantity 500 g
UN Number UN2929
Beilstein 1280477
Solubility Information Soluble in water,alcohols,benzene and acetone.
Formula Weight 96.13
Percent Purity 97%
Chemical Name or Material 2-Cyclohexen-1-one
Afficher plus Afficher moins
compliance-icons compliance-icons
Identifiant produit
  • 2-Cyclohexen-1-one
Mention d'avertissement
  • Danger
Catégorie de danger
  • Acute toxicity Category 2
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
Mention de danger
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H310-Fatal in contact with skin.
  • H319-Causes serious eye irritation.
  • H330-Fatal if inhaled.
Conseils de prudence
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Informations supplémentaires
  • MIXTURE LIST-Contains : 2-Cyclohexene-1-one

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'