missing translation for 'onlineSavingsMsg'
Learn More

2-Cyanobenzeneboronic acid 1,3-propanediol ester, Thermo Scientific™

62.30 € - 208.00 €

Chemical Identifiers

CAS 172732-52-4
Molecular Formula C10H10BNO2
Molecular Weight (g/mol) 187.005
MDL Number MFCD04039012
InChI Key REQZFVYFYAZUMG-UHFFFAOYSA-N
Synonym 2-1,3,2-dioxaborinan-2-yl benzonitrile, 2-cyanophenylboronic acid 1,3-propanediol ester, 2-2-cyanophenyl-1,3,2-dioxaborinane, 2-cyanobenzeneboronic acid 1,3-propanediol ester, 2-cyanophenylboronic acid propanediol ester, 2-cyanophenylboronic acid, 1,3-propanediol cyclic ester, 2-1,3,2-dioxaborinane-2-yl benzonitrile, 2-1,3,2-dioxaborinan-2-yl-benzonitrile, benzonitrile, 2-1,3,2-dioxaborinan-2-yl, 2-cyanophenylboronic acid,1,3-propanediol
PubChem CID 3529602
IUPAC Name 2-(1,3,2-dioxaborinan-2-yl)benzonitrile
SMILES B1(OCCCO1)C2=CC=CC=C2C#N
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15489895
View Documents Promotion Details
Thermo Scientific Alfa Aesar
H52547.MD
250 mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15439287
View Documents Promotion Details
Thermo Scientific Alfa Aesar
H52547.03
1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

172732-52-4
187.005
REQZFVYFYAZUMG-UHFFFAOYSA-N
3529602
B1(OCCCO1)C2=CC=CC=C2C#N
C10H10BNO2
MFCD04039012
2-1,3,2-dioxaborinan-2-yl benzonitrile, 2-cyanophenylboronic acid 1,3-propanediol ester, 2-2-cyanophenyl-1,3,2-dioxaborinane, 2-cyanobenzeneboronic acid 1,3-propanediol ester, 2-cyanophenylboronic acid propanediol ester, 2-cyanophenylboronic acid, 1,3-propanediol cyclic ester, 2-1,3,2-dioxaborinane-2-yl benzonitrile, 2-1,3,2-dioxaborinan-2-yl-benzonitrile, benzonitrile, 2-1,3,2-dioxaborinan-2-yl, 2-cyanophenylboronic acid,1,3-propanediol
2-(1,3,2-dioxaborinan-2-yl)benzonitrile

Specifications

172732-52-4
C10H10BNO2
2-1,3,2-dioxaborinan-2-yl benzonitrile, 2-cyanophenylboronic acid 1,3-propanediol ester, 2-2-cyanophenyl-1,3,2-dioxaborinane, 2-cyanobenzeneboronic acid 1,3-propanediol ester, 2-cyanophenylboronic acid propanediol ester, 2-cyanophenylboronic acid, 1,3-propanediol cyclic ester, 2-1,3,2-dioxaborinane-2-yl benzonitrile, 2-1,3,2-dioxaborinan-2-yl-benzonitrile, benzonitrile, 2-1,3,2-dioxaborinan-2-yl, 2-cyanophenylboronic acid,1,3-propanediol
B1(OCCCO1)C2=CC=CC=C2C#N
187.005
187
42°C to 45°C
MFCD04039012
REQZFVYFYAZUMG-UHFFFAOYSA-N
2-(1,3,2-dioxaborinan-2-yl)benzonitrile
3529602
2-Cyanobenzeneboronic acid 1,3-propanediol ester
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications