missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorothiazole, 97%, Thermo Scientific Chemicals

Product Code. 11409280
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3034-52-4
Molecular Formula C3H2ClNS
Molecular Weight (g/mol) 119.566
MDL Number MFCD00210701
InChI Key KLEYVGWAORGTIT-UHFFFAOYSA-N
Synonym 2-chlorothiazole, thiazole, 2-chloro, 2-thiazolyl chloride, 2-chlothiozole, chlorothiazole, 2-chloro-thiazole, zlchem 495, 2-chlothiazole, pubchem15569
PubChem CID 76429
ChEBI CHEBI:39187
IUPAC Name 2-chloro-1,3-thiazole
SMILES C1=CSC(=N1)Cl

Specifications

Quantity 5 g
Formula Weight 119.57
Percent Purity 97%
Chemical Name or Material 2-Chlorothiazole

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'