missing translation for 'onlineSavingsMsg'
Learn More

2-Chloropropionyl chloride, 95%, Thermo Scientific Chemicals

36.98 € - 394.97 €

Chemical Identifiers

CAS 7623-09-8
Molecular Formula C3H4Cl2O
Molecular Weight (g/mol) 126.97
MDL Number MFCD00000862
InChI Key JEQDSBVHLKBEIZ-UHFFFAOYNA-N
Synonym 2-chloropropionyl chloride, propanoyl chloride, 2-chloro, alpha-chloropropionyl chloride, 2-chloropropionic acid chloride, 2-chloropropionylchloride, .alpha.-chloropropionyl chloride, 2-chloropropanyl chloride, 1,2dichloro-1-propanone, 2-chloro-propionylchloride, 1,2-dichloro-1-propanone
PubChem CID 111019
IUPAC Name 2-chloropropanoyl chloride
SMILES CC(C(=O)Cl)Cl
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10701175
View Documents
Thermo Scientific Acros
295160250
25 mL Glass bottle
36.98 €
25mL
Please to purchase this item. Need a web account? Register with us today!
10023450
View Documents
Thermo Scientific Acros
295161000
100 mL Glass bottle
91.15 €
100mL
Please to purchase this item. Need a web account? Register with us today!
10772961
View Documents
Thermo Scientific Acros
295165000
500 mL Glass bottle
394.97 €
500mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7623-09-8
126.97
JEQDSBVHLKBEIZ-UHFFFAOYNA-N
111019
CC(C(=O)Cl)Cl
C3H4Cl2O
MFCD00000862
2-chloropropionyl chloride, propanoyl chloride, 2-chloro, alpha-chloropropionyl chloride, 2-chloropropionic acid chloride, 2-chloropropionylchloride, .alpha.-chloropropionyl chloride, 2-chloropropanyl chloride, 1,2dichloro-1-propanone, 2-chloro-propionylchloride, 1,2-dichloro-1-propanone
2-chloropropanoyl chloride

Specifications

7623-09-8
100.0
Colorless to Yellow
109.0°C to 111.0°C
Authentic
C3H4Cl2O
1.4390 to 1.4410
02, 248
2-chloropropionyl chloride, propanoyl chloride, 2-chloro, alpha-chloropropionyl chloride, 2-chloropropionic acid chloride, 2-chloropropionylchloride, .alpha.-chloropropionyl chloride, 2-chloropropanyl chloride, 1,2dichloro-1-propanone, 2-chloro-propionylchloride, 1,2-dichloro-1-propanone
JEQDSBVHLKBEIZ-UHFFFAOYNA-N
2-chloropropanoyl chloride
111019
95%
2-Chloropropionyl chloride, 95%
94.0
-71.0°C
1.3000g/mL
34°C
94% min. (GC)
CH3CH(Cl)COCl
MFCD00000862
1.3
Solubility in water: reacts
CC(C(=O)Cl)Cl
126.97
126.97
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
Flammable liquid and vapor.
Contact with water liberates toxic gas.
Reacts violently with water.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or ha

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 231-540-7

missing translation for 'tsca' : TSCA

Product Identifier
  • 2-Chloropropionyl chloride

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 A

Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
  • H226-Flammable liquid and vapour.
  • EUH029-Contact with water liberates toxic gas.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only