missing translation for 'onlineSavingsMsg'
Learn More

2-Chloronicotinamide, 98+%, Thermo Scientific™

14.00 € - 129.00 €

Chemical Identifiers

CAS 10366-35-5
Molecular Formula C6H5ClN2O
Molecular Weight (g/mol) 156.569
MDL Number MFCD00006237
InChI Key ZQZAHPFFZWEUCL-UHFFFAOYSA-N
Synonym 2-chloronicotinamide, 3-pyridinecarboxamide, 2-chloro, chloronicotinamide, 2-chloro-nicotinamide, acmc-1bvtu, nicotinamide, 2-chloro, 2-chloronicotinic acid amide, ksc492e7f, 2-chloro-3-pyridinecarboxamide
PubChem CID 82588
IUPAC Name 2-chloropyridine-3-carboxamide
SMILES C1=CC(=C(N=C1)Cl)C(=O)N
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11480003
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A12090.03
1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11490003
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A12090.06
5 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11400013
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A12090.14
25 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

10366-35-5
156.569
ZQZAHPFFZWEUCL-UHFFFAOYSA-N
82588
C1=CC(=C(N=C1)Cl)C(=O)N
C6H5ClN2O
MFCD00006237
2-chloronicotinamide, 3-pyridinecarboxamide, 2-chloro, chloronicotinamide, 2-chloro-nicotinamide, acmc-1bvtu, nicotinamide, 2-chloro, 2-chloronicotinic acid amide, ksc492e7f, 2-chloro-3-pyridinecarboxamide
2-chloropyridine-3-carboxamide

Specifications

10366-35-5
C6H5ClN2O
119026
ZQZAHPFFZWEUCL-UHFFFAOYSA-N
2-chloropyridine-3-carboxamide
82588
≥98%
163°C to 167°C
MFCD00006237
2-chloronicotinamide, 3-pyridinecarboxamide, 2-chloro, chloronicotinamide, 2-chloro-nicotinamide, acmc-1bvtu, nicotinamide, 2-chloro, 2-chloronicotinic acid amide, ksc492e7f, 2-chloro-3-pyridinecarboxamide
C1=CC(=C(N=C1)Cl)C(=O)N
156.569
156.57
2-Chloronicotinamide
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'einecsNumber' : 233-808-9

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications