missing translation for 'onlineSavingsMsg'
Learn More

2-Chloroisonicotinamide, 95%, Thermo Scientific™

19.30 € - 90.20 €

Chemical Identifiers

CAS 100859-84-5
Molecular Formula C6H5ClN2O
Molecular Weight (g/mol) 156.569
MDL Number MFCD00221401
InChI Key DEMJOLRJLACBRX-UHFFFAOYSA-N
PubChem CID 2800022
IUPAC Name 2-chloropyridine-4-carboxamide
SMILES C1=CN=C(C=C1C(=O)N)Cl
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10243192
View Documents
Thermo Scientific Maybridge
SPB06910DA
1 g Amber Glass Bottle
19.30 €
1g
Please to purchase this item. Need a web account? Register with us today!
10355012
View Documents
Thermo Scientific Maybridge
SPB06910EA
10 g Amber Glass Bottle
90.20 €
10g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

100859-84-5
156.569
DEMJOLRJLACBRX-UHFFFAOYSA-N
2-chloropyridine-4-carboxamide
C6H5ClN2O
MFCD00221401
2800022
C1=CN=C(C=C1C(=O)N)Cl

Specifications

100859-84-5
95%
MFCD00221401
DEMJOLRJLACBRX-UHFFFAOYSA-N
2-chloropyridine-4-carboxamide
2800022
97%
97.0
C6H5ClN2O
2-chloroisonicotinamide, 4-pyridinecarboxamide, 2-chloro, 2-chloro-isonicotinamide, pubchem16074, acmc-1bpcq, 2-chloro-4-pyridinecarboxamide, 4-pyridinecarboxamide,2-chloro
C1=CN=C(C=C1C(=O)N)Cl
156.569
156.57
2-Chloroisonicotinamide
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.