Learn More
2-chloro-6-methyl-4-(trifluoromethyl)pyridine, 97%, Thermo Scientific™
134.00 € - 683.00 €
Chemical Identifiers
CAS | 22123-14-4 |
---|---|
Molecular Formula | C7H5ClF3N |
Molecular Weight (g/mol) | 195.57 |
MDL Number | MFCD00052900 |
InChI Key | SXLBWNSGCIEART-UHFFFAOYSA-N |
Synonym | 2-chloro-6-methyl-4-trifluoromethyl pyridine, 2-chloro-6-methyl-4-trifluoromethyl-pyridine, 2-chlor-6-methyl-4-trifluormethyl pyridine, 2-chloro-4-trifluoromethyl-6-methylpyridine, pyridine, 2-chloro-6-methyl-4-trifluoromethyl, pubchem9219, 6-chloro-4-trifluoromethyl-2-picoline, 2-chloranyl-6-methyl-4-trifluoromethyl pyridine |
PubChem CID | 2736626 |
IUPAC Name | 2-chloro-6-methyl-4-(trifluoromethyl)pyridine |
SMILES | CC1=NC(Cl)=CC(=C1)C(F)(F)F |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10574695
|
Thermo Scientific Maybridge
CD10452DE |
5 g |
134.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10732863
|
Thermo Scientific Maybridge
CD10452EA |
10 g |
201.00 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10634965
|
Thermo Scientific Maybridge
CD10452EE |
50 g | |||||||
Description
Chemical Identifiers
22123-14-4 | |
195.57 | |
SXLBWNSGCIEART-UHFFFAOYSA-N | |
2736626 | |
CC1=NC(Cl)=CC(=C1)C(F)(F)F |
C7H5ClF3N | |
MFCD00052900 | |
2-chloro-6-methyl-4-trifluoromethyl pyridine, 2-chloro-6-methyl-4-trifluoromethyl-pyridine, 2-chlor-6-methyl-4-trifluormethyl pyridine, 2-chloro-4-trifluoromethyl-6-methylpyridine, pyridine, 2-chloro-6-methyl-4-trifluoromethyl, pubchem9219, 6-chloro-4-trifluoromethyl-2-picoline, 2-chloranyl-6-methyl-4-trifluoromethyl pyridine | |
2-chloro-6-methyl-4-(trifluoromethyl)pyridine |
Specifications
22123-14-4 | |
100.0 | |
C7H5ClF3N | |
2-chloro-6-methyl-4-trifluoromethyl pyridine, 2-chloro-6-methyl-4-trifluoromethyl-pyridine, 2-chlor-6-methyl-4-trifluormethyl pyridine, 2-chloro-4-trifluoromethyl-6-methylpyridine, pyridine, 2-chloro-6-methyl-4-trifluoromethyl, pubchem9219, 6-chloro-4-trifluoromethyl-2-picoline, 2-chloranyl-6-methyl-4-trifluoromethyl pyridine | |
CC1=NC(Cl)=CC(=C1)C(F)(F)F | |
195.57 | |
195.57 | |
2-Chloro-6-methyl-4-(trifluoromethyl)pyridine |
96.5 | |
97% | |
MFCD00052900 | |
SXLBWNSGCIEART-UHFFFAOYSA-N | |
2-chloro-6-methyl-4-(trifluoromethyl)pyridine | |
2736626 | |
97% |
Safety and Handling
GHS H Statement:
Harmful if inhaled.
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful in contact with skin.
Causes skin irritation.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Warning