Learn More
2-Chloro-5-fluoropyrimidine, 97%, Thermo Scientific Chemicals
28.00 € - 190.00 €
Chemical Identifiers
CAS | 62802-42-0 |
---|---|
Molecular Formula | C4H2ClFN2 |
Molecular Weight (g/mol) | 132.522 |
MDL Number | MFCD03788197 |
InChI Key | AGYUQBNABXVWMS-UHFFFAOYSA-N |
Synonym | 2-chloro-5-fluoro-pyrimidine, 5-fluoro-2-chloropyrimidine, pyrimidine, 2-chloro-5-fluoro, 2-chlor-5-fluorpyrimidin, 2-chloro-5-fluoro pyrimidine, pubchem4684, acmc-1b6zl, ksc352s4h, #, 2-chloro-5-fluoro-1,3-diazine |
PubChem CID | 583466 |
IUPAC Name | 2-chloro-5-fluoropyrimidine |
SMILES | C1=C(C=NC(=N1)Cl)F |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15461178
|
Thermo Scientific Alfa Aesar
H55055.MD |
250 mg |
28.00 €
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15441178
|
Thermo Scientific Alfa Aesar
H55055.03 |
1 g |
71.70 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15451178
|
Thermo Scientific Alfa Aesar
H55055.06 |
5 g |
190.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
2-Chloro-5-fluoropyrimidine is used to prepare 2,4-disubstituted-5-fluoropyrimidine , which is a biologically active molecule seen in anticancer agents like 5-fluorouracil.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Chloro-5-fluoropyrimidine is used to prepare 2,4-disubstituted-5-fluoropyrimidine , which is a biologically active molecule seen in anticancer agents like 5-fluorouracil.
Notes
Incompatible with strong oxidizing agents.
Chemical Identifiers
62802-42-0 | |
132.522 | |
AGYUQBNABXVWMS-UHFFFAOYSA-N | |
583466 | |
C1=C(C=NC(=N1)Cl)F |
C4H2ClFN2 | |
MFCD03788197 | |
2-chloro-5-fluoro-pyrimidine, 5-fluoro-2-chloropyrimidine, pyrimidine, 2-chloro-5-fluoro, 2-chlor-5-fluorpyrimidin, 2-chloro-5-fluoro pyrimidine, pubchem4684, acmc-1b6zl, ksc352s4h, #, 2-chloro-5-fluoro-1,3-diazine | |
2-chloro-5-fluoropyrimidine |
Specifications
62802-42-0 | |
65°C (149°F) | |
1.503 | |
UN3267 | |
AGYUQBNABXVWMS-UHFFFAOYSA-N | |
2-chloro-5-fluoropyrimidine | |
583466 | |
97% |
1.073 | |
C4H2ClFN2 | |
MFCD03788197 | |
2-chloro-5-fluoro-pyrimidine, 5-fluoro-2-chloropyrimidine, pyrimidine, 2-chloro-5-fluoro, 2-chlor-5-fluorpyrimidin, 2-chloro-5-fluoro pyrimidine, pubchem4684, acmc-1b6zl, ksc352s4h, #, 2-chloro-5-fluoro-1,3-diazine | |
C1=C(C=NC(=N1)Cl)F | |
132.522 | |
132.52 | |
2-Chloro-5-fluoropyrimidine |
Safety and Handling
GHS H Statement
H301-H314-H318-H227
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
P210-P235-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
H227-H302+H332-H314-H335
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, BASIC, ORGANIC, N.O.S.
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only