Learn More
2-Chloro-4-methylpyrimidine, 98%, Thermo Scientific Chemicals
30.04 € - 176.00 €
Chemical Identifiers
CAS | 13036-57-2 |
---|---|
Molecular Formula | C5H5ClN2 |
Molecular Weight (g/mol) | 128.56 |
InChI Key | BHAKRVSCGILCEW-UHFFFAOYSA-N |
Synonym | 2-chloro-4-methyl-pyrimidine, pyrimidine, 2-chloro-4-methyl, 2-chloro-6-methylpyrimidine, pubchem6906, chloro-6 methylpyrimidine, 4-methyl-2-chloropyrimidine, 2-chloro-4-methyl pyrimidine, 4-methyl-2-chloro-pyrimidine, ksc493s3l, 4-methyl-2-pyrimidinylchloride |
PubChem CID | 11629607 |
IUPAC Name | 2-chloro-4-methylpyrimidine |
SMILES | CC1=NC(=NC=C1)Cl |
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
13036-57-2 | |
128.56 | |
2-chloro-4-methyl-pyrimidine, pyrimidine, 2-chloro-4-methyl, 2-chloro-6-methylpyrimidine, pubchem6906, chloro-6 methylpyrimidine, 4-methyl-2-chloropyrimidine, 2-chloro-4-methyl pyrimidine, 4-methyl-2-chloro-pyrimidine, ksc493s3l, 4-methyl-2-pyrimidinylchloride | |
2-chloro-4-methylpyrimidine |
C5H5ClN2 | |
BHAKRVSCGILCEW-UHFFFAOYSA-N | |
11629607 | |
CC1=NC(=NC=C1)Cl |
Specifications
13036-57-2 | |
93°C to 95°C (17.0 mmHg) | |
Glass Bottle | |
2-chloro-4-methyl-pyrimidine, pyrimidine, 2-chloro-4-methyl, 2-chloro-6-methylpyrimidine, pubchem6906, chloro-6 methylpyrimidine, 4-methyl-2-chloropyrimidine, 2-chloro-4-methyl pyrimidine, 4-methyl-2-chloro-pyrimidine, ksc493s3l, 4-methyl-2-pyrimidinylchloride | |
CC1=NC(=NC=C1)Cl | |
128.56 | |
128.56 | |
2-Chloro-4-methylpyrimidine |
51°C to 56°C | |
97.5% min. (GC) | |
C5H5ClN2 | |
BHAKRVSCGILCEW-UHFFFAOYSA-N | |
2-chloro-4-methylpyrimidine | |
11629607 | |
98% |
Safety and Handling
GHS Signal Word: Warning
- 2-Chloro-4-methylpyrimidine, 98%
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.