missing translation for 'onlineSavingsMsg'
Learn More

2'-Chloro-4'-fluoroacetophenone, 98+%, Thermo Scientific™

Product Code. 11416880
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 700-35-6
Molecular Formula C8H6ClFO
Molecular Weight (g/mol) 172.583
MDL Number MFCD00042202
InChI Key CSEMGLVHVZRXQF-UHFFFAOYSA-N
Synonym 1-2-chloro-4-fluorophenyl ethanone, 2'-chloro-4'-fluoroacetophenone, 2-chloro-4-fluoroacetophenone, 1-2-chloro-4-fluorophenyl ethan-1-one, ethanone, 1-2-chloro-4-fluorophenyl, 1-acetyl-2-chloro-4-fluorobenzene, pubchem4187, 2-chloro-4-fluoracetophenone, ksc631o2l, 4'-fluoro-2'-chloroacetophenone
PubChem CID 2736507
IUPAC Name 1-(2-chloro-4-fluorophenyl)ethanone
SMILES CC(=O)C1=C(C=C(C=C1)F)Cl

Specifications

Boiling Point 110°C (19 mmHg)
Refractive Index 1.5255
Quantity 1 g
Beilstein 2249796
Formula Weight 172.59
Percent Purity ≥98%
Chemical Name or Material 2'-Chloro-4'-fluoroacetophenone
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'