missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4,6-diphenyl-1,3,5-triazine, 97%, Thermo Scientific Chemicals

Product Code. 15415977
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

A rational synthesis of kyaphenine (2:4:6-triphenyl-1:3:5-triazine) has been effected by reaction of 2-chloro-4:6-diphenyl-1:3:5-triazine with phenylmagnesium bromide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A rational synthesis of kyaphenine (2:4:6-triphenyl-1:3:5-triazine) has been effected by reaction of 2-chloro-4:6-diphenyl-1:3:5-triazine with phenylmagnesium bromide.

Solubility
Soluble in toluene.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3842-55-5
Molecular Formula C15H10ClN3
Molecular Weight (g/mol) 267.716
MDL Number MFCD05738885
InChI Key DDGPPAMADXTGTN-UHFFFAOYSA-N
Synonym 1,3,5-triazine, 2-chloro-4,6-diphenyl, diphenyl chloro-s-triazine, 4,6-diphenyl-2-chloro-s-triazine, acmc-1ctho, 2-chloro4,6-diphenyl-s-triazine, s-triazine, 2-chloro-4,6-diphenyl, 1,5-triazine, 2-chloro-4,6-diphenyl, 2,6-diphenyl-4-chloro-1,3,5-triazine, 1,3,5-riazine, 2-hloro-4,6-iphenyl, 1,3,5-triazine,2-chloro-4,6-diphenyl
PubChem CID 19698
IUPAC Name 2-chloro-4,6-diphenyl-1,3,5-triazine
SMILES C1=CC=C(C=C1)C2=NC(=NC(=N2)Cl)C3=CC=CC=C3

Specifications

Melting Point 138°C to 140°C
Quantity 25 g
Solubility Information Soluble in toluene.
Formula Weight 267.72
Percent Purity 97%
Chemical Name or Material 2-Chloro-4,6-diphenyl-1,3,5-triazine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.