missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chloro-4,6-diphenyl-1,3,5-triazine, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
A rational synthesis of kyaphenine (2:4:6-triphenyl-1:3:5-triazine) has been effected by reaction of 2-chloro-4:6-diphenyl-1:3:5-triazine with phenylmagnesium bromide.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
A rational synthesis of kyaphenine (2:4:6-triphenyl-1:3:5-triazine) has been effected by reaction of 2-chloro-4:6-diphenyl-1:3:5-triazine with phenylmagnesium bromide.
Solubility
Soluble in toluene.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
A rational synthesis of kyaphenine (2:4:6-triphenyl-1:3:5-triazine) has been effected by reaction of 2-chloro-4:6-diphenyl-1:3:5-triazine with phenylmagnesium bromide.
Solubility
Soluble in toluene.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications
Specifications
Melting Point | 138°C to 140°C |
Quantity | 5 g |
Solubility Information | Soluble in toluene. |
Formula Weight | 267.72 |
Percent Purity | 97% |
Chemical Name or Material | 2-Chloro-4,6-diphenyl-1,3,5-triazine |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction