missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4,6-bis(ethylamino)-1,3,5-triazine, 96%, Thermo Scientific™

Product Code. 11326726
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 122-34-9
Molecular Formula C7H12ClN5
Molecular Weight (g/mol) 201.658
MDL Number MFCD00023174
InChI Key ODCWYMIRDDJXKW-UHFFFAOYSA-N
Synonym simazine, gesatop, princep, simanex, aquazine, batazina, herbazin, symazine, tafazine, taphazine
PubChem CID 5216
ChEBI CHEBI:27496
IUPAC Name 6-chloro-2-N,4-N-diethyl-1,3,5-triazine-2,4-diamine
SMILES CCNC1=NC(=NC(=N1)Cl)NCC

Specifications

Melting Point 227°C to 229°C
Quantity 25 g
UN Number UN3077
Beilstein 10895
Merck Index 14,8533
Formula Weight 201.66
Percent Purity 96%
Chemical Name or Material 2-Chloro-4,6-bis(ethylamino)-1,3,5-triazine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'