Learn More
2'-Chloro-4',5'-difluoroacetophenone, 95%, Thermo Scientific™
30.10 € - 180.00 €
Chemical Identifiers
CAS | 121872-94-4 |
---|---|
Molecular Formula | C8H5ClF2O |
Molecular Weight (g/mol) | 190.574 |
MDL Number | MFCD00671761 |
InChI Key | BNODNMUCMFITCS-UHFFFAOYSA-N |
Synonym | 2'-chloro-4',5'-difluoroacetophenone, 1-2-chloro-4,5-difluorophenyl ethanone, 2-chloro-4,5-difluoroacetophenone, 1-2-chloro-4,5-difluoro-phenyl ethanone, ethanone,1-2-chloro-4,5-difluorophenyl, ethanone, 1-2-chloro-4,5-difluorophenyl, 2-chloro-4 ,5-difluoroacetophenone, 1-acetyl-2-chloro-4,5-difluorobenzene, acmc-20dywg, pubchem4109 |
PubChem CID | 2736489 |
IUPAC Name | 1-(2-chloro-4,5-difluorophenyl)ethanone |
SMILES | CC(=O)C1=CC(=C(C=C1Cl)F)F |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
121872-94-4 | |
190.574 | |
BNODNMUCMFITCS-UHFFFAOYSA-N | |
2736489 | |
CC(=O)C1=CC(=C(C=C1Cl)F)F |
C8H5ClF2O | |
MFCD00671761 | |
2'-chloro-4',5'-difluoroacetophenone, 1-2-chloro-4,5-difluorophenyl ethanone, 2-chloro-4,5-difluoroacetophenone, 1-2-chloro-4,5-difluoro-phenyl ethanone, ethanone,1-2-chloro-4,5-difluorophenyl, ethanone, 1-2-chloro-4,5-difluorophenyl, 2-chloro-4 ,5-difluoroacetophenone, 1-acetyl-2-chloro-4,5-difluorobenzene, acmc-20dywg, pubchem4109 | |
1-(2-chloro-4,5-difluorophenyl)ethanone |
Specifications
121872-94-4 | |
MFCD00671761 | |
BNODNMUCMFITCS-UHFFFAOYSA-N | |
1-(2-chloro-4,5-difluorophenyl)ethanone | |
2736489 | |
95% |
C8H5ClF2O | |
2'-chloro-4',5'-difluoroacetophenone, 1-2-chloro-4,5-difluorophenyl ethanone, 2-chloro-4,5-difluoroacetophenone, 1-2-chloro-4,5-difluoro-phenyl ethanone, ethanone,1-2-chloro-4,5-difluorophenyl, ethanone, 1-2-chloro-4,5-difluorophenyl, 2-chloro-4 ,5-difluoroacetophenone, 1-acetyl-2-chloro-4,5-difluorobenzene, acmc-20dywg, pubchem4109 | |
CC(=O)C1=CC(=C(C=C1Cl)F)F | |
190.574 | |
190.58 | |
2'-Chloro-4',5'-difluoroacetophenone |
Safety and Handling
GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
H315-H319
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
Your input is important to us. Please complete this form to provide feedback related to the content on this product.