missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-1,4-benzoquinone, 95%, Thermo Scientific Chemicals

73.67 € - 292.46 €

Chemical Identifiers

CAS 695-99-8
Molecular Formula C6H3ClO2
Molecular Weight (g/mol) 142.54
MDL Number MFCD00075260
InChI Key WOGWYSWDBYCVDY-UHFFFAOYSA-N
Synonym 2-chloro-1,4-benzoquinone, chloroquinone, monochloroquinone, 2-chloroquinone, chloro-p-benzoquinone, 2-chloro-p-benzoquinone, monochloro-p-benzoquinone, chloro-1,4-benzoquinone, p-benzoquinone, 2-chloro, 2,5-cyclohexadiene-1,4-dione, 2-chloro
PubChem CID 69671
IUPAC Name 2-chlorocyclohexa-2,5-diene-1,4-dione
SMILES ClC1=CC(=O)C=CC1=O
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10653892
View Documents
Thermo Scientific
331220050
5 g Glass bottle
73.67 €
5g
Please to purchase this item. Need a web account? Register with us today!
Currently Unavailable: We have no information when this product will be available
View alternative products
10514141
View Documents
Thermo Scientific Acros
331220250
25 g Glass bottle
292.46 €
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

695-99-8
142.54
WOGWYSWDBYCVDY-UHFFFAOYSA-N
69671
ClC1=CC(=O)C=CC1=O
C6H3ClO2
MFCD00075260
2-chloro-1,4-benzoquinone, chloroquinone, monochloroquinone, 2-chloroquinone, chloro-p-benzoquinone, 2-chloro-p-benzoquinone, monochloro-p-benzoquinone, chloro-1,4-benzoquinone, p-benzoquinone, 2-chloro, 2,5-cyclohexadiene-1,4-dione, 2-chloro
2-chlorocyclohexa-2,5-diene-1,4-dione

Specifications

695-99-8
100.0
Yellow to Brown
95%
MFCD00075260
WOGWYSWDBYCVDY-UHFFFAOYSA-N
2-chlorocyclohexa-2,5-diene-1,4-dione
69671
95%
2-Chloro-1, 4-benzoquinone, 95%
94.0
52.0°C to 57.0°C
Authentic
C6H3ClO2
2-chloro-1,4-benzoquinone, chloroquinone, monochloroquinone, 2-chloroquinone, chloro-p-benzoquinone, 2-chloro-p-benzoquinone, monochloro-p-benzoquinone, chloro-1,4-benzoquinone, p-benzoquinone, 2-chloro, 2,5-cyclohexadiene-1,4-dione, 2-chloro
ClC1=CC(=O)C=CC1=O
142.54
142.54
Crystals
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if inhaled.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

missing translation for 'tsca' : TSCA

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.