missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-1,3-thiazole-5-carbonyl chloride, 97%, Thermo Scientific™

Product Code. 10722944
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 148637-74-5
Molecular Formula C4HCl2NOS
Molecular Weight (g/mol) 182.018
MDL Number MFCD09879938
InChI Key GZSJPOBGZMUSPV-UHFFFAOYSA-N
Synonym 5-thiazolecarbonylchloride, 2-chloro, acmc-20n5hc, 2-chlorothiazole-5-carbonylchloride, 2-chlorothiazole-5-carbonyl chloride, 2-chloro-thiazole-5-carbonyl chloride, 5-thiazolecarbonyl chloride, 2-chloro, 5-thiazolecarbonyl chloride, 2-chloro-9ci
PubChem CID 24229701
IUPAC Name 2-chloro-1,3-thiazole-5-carbonyl chloride
SMILES C1=C(SC(=N1)Cl)C(=O)Cl

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 182.03
Percent Purity 97%
Chemical Name or Material 2-Chloro-1, 3-thiazole-5-carbonyl chloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'