missing translation for 'onlineSavingsMsg'
Learn More

2-Carboxybenzeneboronic acid, 95%, Thermo Scientific Chemicals

Product Code. 11322977
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 149105-19-1
Molecular Formula C7H7BO4
Molecular Weight (g/mol) 165.939
MDL Number MFCD01318118
InChI Key KWNPRVWFJOSGMZ-UHFFFAOYSA-N
Synonym 2-carboxyphenylboronic acid, 2-dihydroxyboranyl benzoic acid, 2-carboxybenzeneboronic acid, 2-dihydroxyboronyl benzoic acid, 2-dihydroxyboryl benzoic acid, carboxyphenylboronic acid, benzoic acid, 2-borono, o-carboxyphenylboronic acid, 2-carboxyphenylboronicacid, borobenzoesaure
PubChem CID 2733985
IUPAC Name 2-boronobenzoic acid
SMILES B(C1=CC=CC=C1C(=O)O)(O)O

Specifications

Melting Point ∼220°C
Quantity 1 g
Beilstein 2835407
Formula Weight 165.94
Percent Purity 95%
Chemical Name or Material 2-Carboxybenzeneboronic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'