missing translation for 'onlineSavingsMsg'
Learn More

2-Butyn-1-ol, 98%, Thermo Scientific Chemicals

52.86 € - 673.35 €

Chemical Identifiers

CAS 764-01-2
Molecular Formula C4H6O
Molecular Weight (g/mol) 70.091
MDL Number MFCD00002914
InChI Key NEEDEQSZOUAJMU-UHFFFAOYSA-N
Synonym 2-butyn-1-ol, 2-butynol, 2-butyne-1-ol, hydroxymethylmethylacetylene, 2-butynyl alcohol, 1-hydroxybut-2-yne, 2-butyn-l-ol, 2-butyn1-ol, 2-butin-1-ol, but-2-in-1-ol
PubChem CID 12991
IUPAC Name but-2-yn-1-ol
SMILES CC#CCO
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11457677
View Documents
Thermo Scientific Alfa Aesar
A15709.06
5 g
52.86 €
5g
Please to purchase this item. Need a web account? Register with us today!
11467677
View Documents
Thermo Scientific Alfa Aesar
A15709.14
25 g
212.06 €
25g
Please to purchase this item. Need a web account? Register with us today!
11477677
View Documents
Thermo Scientific Alfa Aesar
A15709.22
100 g
673.35 €
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

2-Butyn-1-ol is used in the synthesis of homocoupling products. It is also used to inhibit corrosion of iron in acidic solution.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Butyn-1-ol is used in the synthesis of homocoupling products. It is also used to inhibit corrosion of iron in acidic solution.

Solubility
Miscible with chloroform and methanol.

Notes
Incompatible with oxidizing agents, acids, acid anhydrides and copper.
Specifications

Chemical Identifiers

764-01-2
70.091
NEEDEQSZOUAJMU-UHFFFAOYSA-N
12991
CC#CCO
C4H6O
MFCD00002914
2-butyn-1-ol, 2-butynol, 2-butyne-1-ol, hydroxymethylmethylacetylene, 2-butynyl alcohol, 1-hydroxybut-2-yne, 2-butyn-l-ol, 2-butyn1-ol, 2-butin-1-ol, but-2-in-1-ol
but-2-yn-1-ol

Specifications

764-01-2
0.938
51°C (123°F)
C4H6O
MFCD00002914
1733676
Miscible with chloroform and methanol.
CC#CCO
70.091
70.09
2-Butyn-1-ol
-2°C
142°C to 143°C
Characteristic
1.453
UN1987
2-butyn-1-ol, 2-butynol, 2-butyne-1-ol, hydroxymethylmethylacetylene, 2-butynyl alcohol, 1-hydroxybut-2-yne, 2-butyn-l-ol, 2-butyn1-ol, 2-butin-1-ol, but-2-in-1-ol
NEEDEQSZOUAJMU-UHFFFAOYSA-N
but-2-yn-1-ol
12991
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c

H226-H315-H319-H335

missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALCOHOLS, N.O.S.

missing translation for 'einecsNumber' : 212-113-4

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

Product Identifier
  • 2-Butyn-1-ol

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H226-Flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

Supplemental information
  • MIXTURE LIST-Contains : But-2-yn-1-ol

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only