missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Butanone, ReagentPlus™, ≥99%, Honeywell™
Reagent Grade,≥99%
47.30 € - 670.45 €
Specifications
Melting Point | -87°C |
---|---|
Appearance | Colorless |
Density | 0.805g/mL at 25°C |
Boiling Point | 79°C to 81°C |
Flash Point | −3°C (26.6°F) |
Refractive Index | n20/D 1.379(lit.) |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15621010
|
Honeywell Research Chemicals
110264-500ML |
500 mL | Glass Bottle | N/A | |||||
15681000
|
Honeywell Research Chemicals
110264-1L |
1 L | Glass Bottle | N/A | |||||
15691000
|
Honeywell Research Chemicals
110264-2.5L |
2.5 L | Glass Bottle | N/A | |||||
15611010
|
Honeywell Research Chemicals
110264-4X2.5L |
4 x 2.5 L | Glass Bottle | N/A | |||||
15601010
|
Honeywell Research Chemicals
110264-25L |
25 L | Stainless Steel Can | N/A | |||||
Description
- ≥95% for general laboratory use.
Specifications
-87°C | |
0.805g/mL at 25°C | |
−3°C (26.6°F) |
Colorless | |
79°C to 81°C | |
n20/D 1.379(lit.) |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title