missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Butanol, puriss. p.a., Reag. Ph. Eur., ≥99.5% (GC), Honeywell Riedel-de Haën™
Puriss. p.a., Reag. Ph. Eur.,≥99.5% (GC)
29.87 € - 185.25 €
Chemical Identifiers
CAS | 78-92-2 |
---|---|
Molecular Formula | C4H10O |
Molecular Weight (g/mol) | 74.12 |
MDL Number | MFCD00004569 |
InChI Key | BTANRVKWQNVYAZ-UHFFFAOYNA-N |
Synonym | 2-butanol, sec-butanol, sec-butyl alcohol, 2-hydroxybutane, 2-butyl alcohol, s-butyl alcohol, butylene hydrate, s-butanol, 1-methyl-1-propanol, 1-methylpropyl alcohol |
PubChem CID | 6568 |
ChEBI | CHEBI:35687 |
IUPAC Name | butan-2-ol |
SMILES | CCC(C)O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15691700
|
Honeywell-Riedel-de Haen
19440-250ML |
250 mL | Glass Bottle | N/A | |||||
15671700
|
Honeywell-Riedel-de Haen
19440-1L |
1 L | Glass Bottle |
97.42 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15681700
|
Honeywell-Riedel-de Haen
19440-2.5L |
2.5 L | Glass Bottle |
185.25 €
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Honeywell Riedel-de Haen has over 200 years of history in producing high-purity chemicals
- It is the leading brand offering High-Purity Solvents for dedicated applications.
Chemical Identifiers
78-92-2 | |
74.12 | |
BTANRVKWQNVYAZ-UHFFFAOYNA-N | |
6568 | |
butan-2-ol |
C4H10O | |
MFCD00004569 | |
2-butanol, sec-butanol, sec-butyl alcohol, 2-hydroxybutane, 2-butyl alcohol, s-butyl alcohol, butylene hydrate, s-butanol, 1-methyl-1-propanol, 1-methylpropyl alcohol | |
CHEBI:35687 | |
CCC(C)O |
Specifications
-115°C | |
Colorless | |
99°C | |
27°C (80.6°F) | |
n20/D 1.397 | |
CH3CH2CH(OH)CH3 | |
12.5mmHg (20°C) | |
773649 | |
BTANRVKWQNVYAZ-UHFFFAOYNA-N | |
butan-2-ol | |
6568 | |
74.12g/mol | |
puriss. p.a. |
78-92-2 | |
0.808g/mL at 25°C | |
0.098 | |
Liquid | |
C4H10O | |
MFCD00004569 | |
UN1120 | |
2-butanol, sec-butanol, sec-butyl alcohol, 2-hydroxybutane, 2-butyl alcohol, s-butyl alcohol, butylene hydrate, s-butanol, 1-methyl-1-propanol, 1-methylpropyl alcohol | |
CCC(C)O | |
74.12 | |
CHEBI:35687 | |
≥99.5% (GC) | |
2-Butanol, Reag. Ph. Eur. |
Safety and Handling
P210-P305 + P351 + P338-P370 + P378
H226-H319-H335-H336
missing translation for 'dotInformation' : Class 3 - PG 3 - Butanols
missing translation for 'einecsNumber' : 201-158-5
missing translation for 'rtecsNumber' : EO1750000
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title