Learn More
2-Bromothiazole, 99%, Thermo Scientific Chemicals
77.80 € - 1212.00 €
Chemical Identifiers
CAS | 3034-53-5 |
---|---|
Molecular Formula | C3H2BrNS |
Molecular Weight (g/mol) | 164.02 |
MDL Number | MFCD00005316 |
InChI Key | RXNZFHIEDZEUQM-UHFFFAOYSA-N |
Synonym | 2-bromothiazole, thiazole, 2-bromo, 2-bromo thiazole, 2-bromo-thiazole, 2-thiazolyl bromide, thiazole, bromo, bromothiazole, bromo-thiazole, 2 bromothiazole, 2-bromothiazol |
PubChem CID | 76430 |
IUPAC Name | 2-bromo-1,3-thiazole |
SMILES | C1=CSC(=N1)Br |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11432607
|
Thermo Scientific Alfa Aesar
A14838.09 |
10 g |
77.80 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11319659
|
Thermo Scientific Alfa Aesar
A14838.14 |
25 g |
165.00 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11442607
|
Thermo Scientific Alfa Aesar
A14838.18 |
50 g |
269.00 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11452607
|
Thermo Scientific Alfa Aesar
A14838.30 |
250 g | |||||||
Description
2-Bromothiazole, is intended used for research purpose only. It is also used as Aryl halide which are used to N-arylate 5- and 7-azaindoles. Copper-catalyzed cyanation provides 2-cyanothiazole
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Bromothiazole, is intended used for research purpose only. It is also used as Aryl halide which are used to N-arylate 5- and 7-azaindoles. Copper-catalyzed cyanation provides 2-cyanothiazole
Solubility
It is insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from Strong oxidizing agents, Hydrogen bromide gas, nitrogen oxides (NOx), Sulphur oxides.
Chemical Identifiers
3034-53-5 | |
164.02 | |
RXNZFHIEDZEUQM-UHFFFAOYSA-N | |
76430 | |
C1=CSC(=N1)Br |
C3H2BrNS | |
MFCD00005316 | |
2-bromothiazole, thiazole, 2-bromo, 2-bromo thiazole, 2-bromo-thiazole, 2-thiazolyl bromide, thiazole, bromo, bromothiazole, bromo-thiazole, 2 bromothiazole, 2-bromothiazol | |
2-bromo-1,3-thiazole |
Specifications
3034-53-5 | |
171°C | |
C3H2BrNS | |
MFCD00005316 | |
2-bromothiazole, thiazole, 2-bromo, 2-bromo thiazole, 2-bromo-thiazole, 2-thiazolyl bromide, thiazole, bromo, bromothiazole, bromo-thiazole, 2 bromothiazole, 2-bromothiazol | |
RXNZFHIEDZEUQM-UHFFFAOYSA-N | |
2-bromo-1,3-thiazole | |
76430 | |
99% |
1.836 | |
63°C (145°F) | |
1.593 | |
105724 | |
It is insoluble in water. | |
C1=CSC(=N1)Br | |
164.02 | |
164.02 | |
2-Bromothiazole |
Safety and Handling
GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
H227-H315-H319-H335
missing translation for 'einecsNumber' : 221-229-4
missing translation for 'tsca' : Yes
Recommended Storage : Keep cold
RUO – Research Use Only