Learn More
2-Bromopentane, tech. 90%, Thermo Scientific Chemicals
38.70 € - 337.00 €
Chemical Identifiers
CAS | 107-81-3 |
---|---|
Molecular Formula | C5H11Br |
Molecular Weight (g/mol) | 151.047 |
MDL Number | MFCD00000160 |
InChI Key | LGAJYTCRJPCZRJ-UHFFFAOYSA-N |
Synonym | pentane, 2-bromo, 2-pentyl bromide, 1-methylbutyl bromide, +/--2-bromopentane, sec-amyl bromide, 2-bromo-pentane, 2-bromanylpentane, 2-bromopentane, 4-01-00-00312 beilstein handbook reference, ksc176i8j |
PubChem CID | 7890 |
IUPAC Name | 2-bromopentane |
SMILES | CCCC(C)Br |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11403427
|
Thermo Scientific Alfa Aesar
A17205.14 |
25 g |
38.70 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11413427
|
Thermo Scientific Alfa Aesar
A17205.22 |
100 g |
89.20 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11423427
|
Thermo Scientific Alfa Aesar
A17205.36 |
500 g |
337.00 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Used as a chemical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsUsed as a chemical intermediate.
Solubility
Not miscible or difficult to mix in water. Soluble in Dichloromethane, Ether, Ethyl Acetate, Hexane and Methanol
Notes
Store away from oxidizing agents. Keep the container tightly sealed and store in cool, dry condition in well ventilated place.
Chemical Identifiers
107-81-3 | |
151.047 | |
LGAJYTCRJPCZRJ-UHFFFAOYSA-N | |
7890 | |
CCCC(C)Br |
C5H11Br | |
MFCD00000160 | |
pentane, 2-bromo, 2-pentyl bromide, 1-methylbutyl bromide, +/--2-bromopentane, sec-amyl bromide, 2-bromo-pentane, 2-bromanylpentane, 2-bromopentane, 4-01-00-00312 beilstein handbook reference, ksc176i8j | |
2-bromopentane |
Specifications
107-81-3 | |
117°C to 118°C | |
Characteristic | |
MFCD00000160 | |
1718829 | |
LGAJYTCRJPCZRJ-UHFFFAOYSA-N | |
2-bromopentane | |
7890 | |
90% | |
2-Bromopentane |
-96°C | |
20°C (68°F) | |
C5H11Br | |
UN2343 | |
pentane, 2-bromo, 2-pentyl bromide, 1-methylbutyl bromide, +/--2-bromopentane, sec-amyl bromide, 2-bromo-pentane, 2-bromanylpentane, 2-bromopentane, 4-01-00-00312 beilstein handbook reference, ksc176i8j | |
CCCC(C)Br | |
151.047 | |
151.05 | |
Technical |
Safety and Handling
GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P210-P280g-P273-P305+P351+P338
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves.
Avoid release to the environment.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Danger
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: 2-BROMOPENTANE
missing translation for 'rtecsNumber' : RZ9800000
missing translation for 'tsca' : TSCA
RUO – Research Use Only