Learn More
2-(Bromomethyl)benzothiazole, 98+%, Thermo Scientific Chemicals
45.40 € - 107.00 €
Chemical Identifiers
CAS | 106086-78-6 |
---|---|
Molecular Formula | C8H6BrNS |
Molecular Weight (g/mol) | 228.107 |
MDL Number | MFCD00461195 |
InChI Key | WFLCAOGKZQTOIG-UHFFFAOYSA-N |
Synonym | 2-bromomethyl-1,3-benzothiazole, 2-bromomethyl benzo d thiazole, 2-bromomethyl benzothiazole, benzothiazole, 2-bromomethyl, benzo d thiazol-2-yl methyl bromide, zlchem 58, pubchem3912, acmc-1bpju, 2-bromomethylbenzothiazole |
PubChem CID | 2776258 |
IUPAC Name | 2-(bromomethyl)-1,3-benzothiazole |
SMILES | C1=CC=C2C(=C1)N=C(S2)CBr |
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
106086-78-6 | |
228.107 | |
WFLCAOGKZQTOIG-UHFFFAOYSA-N | |
2776258 | |
C1=CC=C2C(=C1)N=C(S2)CBr |
C8H6BrNS | |
MFCD00461195 | |
2-bromomethyl-1,3-benzothiazole, 2-bromomethyl benzo d thiazole, 2-bromomethyl benzothiazole, benzothiazole, 2-bromomethyl, benzo d thiazol-2-yl methyl bromide, zlchem 58, pubchem3912, acmc-1bpju, 2-bromomethylbenzothiazole | |
2-(bromomethyl)-1,3-benzothiazole |
Specifications
106086-78-6 | |
C8H6BrNS | |
UN3261 | |
WFLCAOGKZQTOIG-UHFFFAOYSA-N | |
2-(bromomethyl)-1,3-benzothiazole | |
2776258 | |
≥98% |
47°C to 49°C | |
MFCD00461195 | |
2-bromomethyl-1,3-benzothiazole, 2-bromomethyl benzo d thiazole, 2-bromomethyl benzothiazole, benzothiazole, 2-bromomethyl, benzo d thiazol-2-yl methyl bromide, zlchem 58, pubchem3912, acmc-1bpju, 2-bromomethylbenzothiazole | |
C1=CC=C2C(=C1)N=C(S2)CBr | |
228.107 | |
228.11 | |
2-(Bromomethyl)benzothiazole |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
missing translation for 'tsca' : No
Recommended Storage : Keep cold
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.