Learn More
2-(Bromomethyl)-1-methyl-1H-benzimidazole hydrobromide, 97%, Thermo Scientific™
96.60 € - 111.00 €
Chemical Identifiers
CAS | 934570-40-8 |
---|---|
Molecular Formula | C9H10Br2N2 |
Molecular Weight (g/mol) | 306.001 |
MDL Number | MFCD09702343 |
InChI Key | WHMCWOHHFOGLGP-UHFFFAOYSA-N |
Synonym | 2-bromomethyl-1-methyl-1h-benzimidazole hydrobromide, 2-bromomethyl-1-methyl-1h-benzo d imidazole hydrobromide, 2-bromomethyl-1-methyl-1,3-benzodiazole hydrobromide, 2-bromomethyl-1-methyl-benzimidazole hydrobromide, 2-bromomethyl-1-methyl-1h-benzimidazole hbr, 2-bromomethyl-1-methyl-1h-benzimidazole-hydrogen bromide 1/1 |
PubChem CID | 24229451 |
IUPAC Name | 2-(bromomethyl)-1-methylbenzimidazole;hydrobromide |
SMILES | CN1C2=CC=CC=C2N=C1CBr.Br |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10003583
|
Thermo Scientific Maybridge
CC03858CB |
250 mg | Amber Glass Bottle |
96.60 €
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10626994
|
Thermo Scientific Maybridge
CC03858DA |
1 g | Amber Glass Bottle |
111.00 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Chemical Identifiers
934570-40-8 | |
306.001 | |
WHMCWOHHFOGLGP-UHFFFAOYSA-N | |
24229451 | |
CN1C2=CC=CC=C2N=C1CBr.Br |
C9H10Br2N2 | |
MFCD09702343 | |
2-bromomethyl-1-methyl-1h-benzimidazole hydrobromide, 2-bromomethyl-1-methyl-1h-benzo d imidazole hydrobromide, 2-bromomethyl-1-methyl-1,3-benzodiazole hydrobromide, 2-bromomethyl-1-methyl-benzimidazole hydrobromide, 2-bromomethyl-1-methyl-1h-benzimidazole hbr, 2-bromomethyl-1-methyl-1h-benzimidazole-hydrogen bromide 1/1 | |
2-(bromomethyl)-1-methylbenzimidazole;hydrobromide |
Specifications
934570-40-8 | |
100.0 | |
C9H10Br2N2 | |
2-bromomethyl-1-methyl-1h-benzimidazole hydrobromide, 2-bromomethyl-1-methyl-1h-benzo d imidazole hydrobromide, 2-bromomethyl-1-methyl-1,3-benzodiazole hydrobromide, 2-bromomethyl-1-methyl-benzimidazole hydrobromide, 2-bromomethyl-1-methyl-1h-benzimidazole hbr, 2-bromomethyl-1-methyl-1h-benzimidazole-hydrogen bromide 1/1 | |
CN1C2=CC=CC=C2N=C1CBr.Br | |
306.001 | |
306 | |
2-(Bromomethyl)-1-methyl-1H-benzimidazole hydrobromide |
96.5 | |
97% | |
MFCD09702343 | |
WHMCWOHHFOGLGP-UHFFFAOYSA-N | |
2-(bromomethyl)-1-methylbenzimidazole;hydrobromide | |
24229451 | |
97% |
Safety and Handling
GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Harmful if swallowed.
Causes serious eye irritation.
Harmful in contact with skin.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning