Learn More
(2-Bromoethyl)cyclohexane, 99%, Thermo Scientific Chemicals
26.60 € - 278.00 €
Chemical Identifiers
CAS | 1647-26-3 |
---|---|
Molecular Formula | C8H15Br |
Molecular Weight (g/mol) | 191.11 |
MDL Number | MFCD00019398 |
InChI Key | JRQAAYVLPPGEHT-UHFFFAOYSA-N |
Synonym | 2-bromoethyl cyclohexane, 2-cyclohexylethyl bromide, cyclohexane, 2-bromoethyl, 2-cyclohexylethylbromide, 1-bromo-2-cyclohexylethane, cyclohexylethylbromide, bromoethyl cyclohexane, cyclohexylethyl bromide, acmc-1bzct, 1-bromo-2cyclohexylethane |
PubChem CID | 15440 |
IUPAC Name | 2-bromoethylcyclohexane |
SMILES | BrCCC1CCCCC1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11377436
|
Thermo Scientific Alfa Aesar
L09466.03 |
1 g |
26.60 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11387436
|
Thermo Scientific Alfa Aesar
L09466.06 |
5 g |
74.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11397436
|
Thermo Scientific Alfa Aesar
L09466.14 |
25 g |
278.00 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1647-26-3 | |
191.11 | |
JRQAAYVLPPGEHT-UHFFFAOYSA-N | |
15440 | |
BrCCC1CCCCC1 |
C8H15Br | |
MFCD00019398 | |
2-bromoethyl cyclohexane, 2-cyclohexylethyl bromide, cyclohexane, 2-bromoethyl, 2-cyclohexylethylbromide, 1-bromo-2-cyclohexylethane, cyclohexylethylbromide, bromoethyl cyclohexane, cyclohexylethyl bromide, acmc-1bzct, 1-bromo-2cyclohexylethane | |
2-bromoethylcyclohexane |
Specifications
1647-26-3 | |
70°C to 71°C (6 mmHg) | |
C8H15Br | |
MFCD00019398 | |
2-bromoethyl cyclohexane, 2-cyclohexylethyl bromide, cyclohexane, 2-bromoethyl, 2-cyclohexylethylbromide, 1-bromo-2-cyclohexylethane, cyclohexylethylbromide, bromoethyl cyclohexane, cyclohexylethyl bromide, acmc-1bzct, 1-bromo-2cyclohexylethane | |
BrCCC1CCCCC1 | |
191.11 | |
191.12 | |
(2-Bromoethyl)cyclohexane |
1.221 | |
96°C (204°F) | |
1.49 | |
1919816 | |
JRQAAYVLPPGEHT-UHFFFAOYSA-N | |
2-bromoethylcyclohexane | |
15440 | |
99% |
Safety and Handling
GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 216-712-1
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
- (2-Bromoethyl)cyclohexane
Signal Word
- Warning
Hazard Category
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P362-Take off contaminated clothing.
- P321-Specific treatment (see on this label).
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.