missing translation for 'onlineSavingsMsg'
Learn More

(2-Bromoethyl)benzene, 98%, Thermo Scientific Chemicals

40.00 € - 402.00 €

Chemical Identifiers

CAS 103-63-9
MDL Number MFCD00000240
InChI Key WMPPDTMATNBGJN-UHFFFAOYSA-N
Synonym 2-bromoethyl benzene, phenethyl bromide, 2-phenylethyl bromide, 1-bromo-2-phenylethane, phenylethyl bromide, benzene, 2-bromoethyl, 2-phenethyl bromide, beta-bromoethylbenzene, 2-phenyl-1-bromoethane, phenethylbromide
PubChem CID 7666
IUPAC Name 2-bromoethylbenzene
SMILES C1=CC=C(C=C1)CCBr
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10000660
View Documents Promotion Details
Thermo Scientific Acros
106941000
100 mL Glass bottle
40.00 €
100mL
Please to purchase this item. Need a web account? Register with us today!
10569810
View Documents Promotion Details
Thermo Scientific Acros
106945000
500 mL Glass bottle
102.51 €
500mL
Please to purchase this item. Need a web account? Register with us today!
10319093
View Documents Promotion Details
Thermo Scientific Acros
106940025
2.5 L Glass bottle
402.00 €
2.50 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

103-63-9
WMPPDTMATNBGJN-UHFFFAOYSA-N
7666
C1=CC=C(C=C1)CCBr
MFCD00000240
2-bromoethyl benzene, phenethyl bromide, 2-phenylethyl bromide, 1-bromo-2-phenylethane, phenylethyl bromide, benzene, 2-bromoethyl, 2-phenethyl bromide, beta-bromoethylbenzene, 2-phenyl-1-bromoethane, phenethylbromide
2-bromoethylbenzene

Specifications

103-63-9
100.0
Colorless to Yellow
220°C to 221°C
Authentic
C8H9Br
1.5553 to 1.5573
05,356
2-bromoethyl benzene, phenethyl bromide, 2-phenylethyl bromide, 1-bromo-2-phenylethane, phenylethyl bromide, benzene, 2-bromoethyl, 2-phenethyl bromide, beta-bromoethylbenzene, 2-phenyl-1-bromoethane, phenethylbromide
WMPPDTMATNBGJN-UHFFFAOYSA-N
2-bromoethylbenzene
7666
98%
(2-Bromoethyl)benzene, 98%
97.5
-56°C
1.3500g/mL
89°C
97.5% min. (GC)
C6H5(CH2)2Br
MFCD00000240
1.35
Solubility in water: insoluble. Other solubilities: soluble in ether and benzene
C1=CC=C(C=C1)CCBr
185.06
185.06
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Causes serious eye irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

GHS Signal Word: Warning

missing translation for 'einecsNumber' : 203-130-8

missing translation for 'rtecsNumber' : CY9032000

missing translation for 'tsca' : TSCA

Product Identifier
  • (2-Bromoethyl)benzene

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2

Hazard Statement
  • H302-Harmful if swallowed.
  • H319-Causes serious eye irritation.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only