missing translation for 'onlineSavingsMsg'
Learn More

(2-Bromoethoxy)-tert-butyldimethylsilane, 98%, stab. with sodium carbonate, Thermo Scientific Chemicals

33.65 € - 106.00 €

Chemical Identifiers

CAS 86864-60-0
Molecular Formula C8H19BrOSi
Molecular Weight (g/mol) 239.228
MDL Number MFCD00209550
InChI Key JBKINHFZTVLNEM-UHFFFAOYSA-N
Synonym 2-bromoethoxy-tert-butyldimethylsilane, 2-bromoethoxy tert-butyl dimethylsilane, 2-bromoethoxy-t-butyl dimethylsilane, 2-t-butyldimethylsiloxy ethylbromide, 2-bromoethoxy-t-butyldimethylsilane, 2-bromo-ethoxy-tert-butyl-dimethyl-silane, silane, 2-bromoethoxy 1,1-dimethylethyl dimethyl, brch2ch2otbs
PubChem CID 3608067
IUPAC Name 2-bromoethoxy-tert-butyl-dimethylsilane
SMILES CC(C)(C)[Si](C)(C)OCCBr
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15472786
View Documents
Thermo Scientific Alfa Aesar
H56594.03
1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15482786
View Documents
Thermo Scientific Alfa Aesar
H56594.06
5 g
106.00 €
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

86864-60-0
239.228
JBKINHFZTVLNEM-UHFFFAOYSA-N
3608067
CC(C)(C)[Si](C)(C)OCCBr
C8H19BrOSi
MFCD00209550
2-bromoethoxy-tert-butyldimethylsilane, 2-bromoethoxy tert-butyl dimethylsilane, 2-bromoethoxy-t-butyl dimethylsilane, 2-t-butyldimethylsiloxy ethylbromide, 2-bromoethoxy-t-butyldimethylsilane, 2-bromo-ethoxy-tert-butyl-dimethyl-silane, silane, 2-bromoethoxy 1,1-dimethylethyl dimethyl, brch2ch2otbs
2-bromoethoxy-tert-butyl-dimethylsilane

Specifications

120°C (100mmHg)
Colorless
C8H19BrOSi
73°C (163°F)
JBKINHFZTVLNEM-UHFFFAOYSA-N
2-bromoethoxy-tert-butyl-dimethylsilane
3608067
98%
1.115 g/mL
86864-60-0
Liquid
MFCD00209550
2-bromoethoxy-tert-butyldimethylsilane, 2-bromoethoxy tert-butyl dimethylsilane, 2-bromoethoxy-t-butyl dimethylsilane, 2-t-butyldimethylsiloxy ethylbromide, 2-bromoethoxy-t-butyldimethylsilane, 2-bromo-ethoxy-tert-butyl-dimethyl-silane, silane, 2-bromoethoxy 1,1-dimethylethyl dimethyl, brch2ch2otbs
CC(C)(C)[Si](C)(C)OCCBr
239.228
239.23
1.444
(2-Bromoethoxy)-tert-butyldimethylsilane, Stabilized with sodium carbonate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.

P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c

H227-H315-H319-H335

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.