missing translation for 'onlineSavingsMsg'
Learn More

2-Bromoethanol, 97%, Thermo Scientific Chemicals

30.75 € - 477.38 €

Chemical Identifiers

CAS 540-51-2
Molecular Formula C2H5BrO
Molecular Weight (g/mol) 124.965
MDL Number MFCD00002827
InChI Key LDLCZOVUSADOIV-UHFFFAOYSA-N
Synonym ethylene bromohydrin, ethanol, 2-bromo, 2-bromoethan-1-ol, glycol bromohydrin, 2-hydroxyethyl bromide, 2-bromo-1-ethanol, 1-bromo-2-ethanol, ethylenebromohydrin, 2-bromoethyl alcohol, 2-bromo-ethanol
PubChem CID 10898
IUPAC Name 2-bromoethanol
SMILES C(CBr)O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11329669
View Documents
Thermo Scientific Alfa Aesar
A10275.14
25 g
30.75 €
25g
Please to purchase this item. Need a web account? Register with us today!
11464003
View Documents
Thermo Scientific Alfa Aesar
A10275.30
250 g
137.69 €
250g
Please to purchase this item. Need a web account? Register with us today!
11454003
View Documents
Thermo Scientific Alfa Aesar
A10275.0B
1000 g
477.38 €
1000g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

2-Bromoethanol is utilized in selective reduction of nitroarenes. It is used as a solvent and organic synthetic raw material. It finds application in the synthesis of 2-piperidin-1-yl-ethanol by reacting with piperidine. It is used in selective reduction of nitroarenes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromoethanol is utilized in selective reduction of nitroarenes. It is used as a solvent and organic synthetic raw material. It finds application in the synthesis of 2-piperidin-1-yl-ethanol by reacting with piperidine. It is used in selective reduction of nitroarenes.

Solubility
Miscible with water, organic solvent and petroleum ether.

Notes
Light sensitive. Hygroscopic. Store in a cool place. Incompatible with strong acids, strong oxidizing agents, strong reducing agents, acid chlorides and acid anhydrides.
Specifications

Chemical Identifiers

540-51-2
124.965
LDLCZOVUSADOIV-UHFFFAOYSA-N
10898
C(CBr)O
C2H5BrO
MFCD00002827
ethylene bromohydrin, ethanol, 2-bromo, 2-bromoethan-1-ol, glycol bromohydrin, 2-hydroxyethyl bromide, 2-bromo-1-ethanol, 1-bromo-2-ethanol, ethylenebromohydrin, 2-bromoethyl alcohol, 2-bromo-ethanol
2-bromoethanol

Specifications

540-51-2
1.762
>110°C (230°F)
BrCH2CH2OH
MFCD00002827
878140
14,3792
Miscible with water,organic solvent and petroleum ether.
C(CBr)O
124.965
124.97
2-Bromoethanol
-80°C
149°C to 150°C
C2H5BrO
1.493
UN2922
Hygroscopic; Light sensitive
ethylene bromohydrin, ethanol, 2-bromo, 2-bromoethan-1-ol, glycol bromohydrin, 2-hydroxyethyl bromide, 2-bromo-1-ethanol, 1-bromo-2-ethanol, ethylenebromohydrin, 2-bromoethyl alcohol, 2-bromo-ethanol
LDLCZOVUSADOIV-UHFFFAOYSA-N
2-bromoethanol
10898
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H300-H310-H330-H314-H318
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P262-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c

H301+H331-H310-H314

missing translation for 'dotInformation' : Hazard Class: 6.1; Packaging Group: II

missing translation for 'einecsNumber' : 208-748-1

missing translation for 'rtecsNumber' : KJ8225000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H331-Toxic if inhaled.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.