missing translation for 'onlineSavingsMsg'
Learn More

2-Bromoacetophenone, 98%, Thermo Scientific Chemicals

32.86 € - 694.00 €

Chemical Identifiers

CAS 70-11-1
Molecular Formula C8H7BrO
Molecular Weight (g/mol) 199.05
MDL Number MFCD00000195
InChI Key LIGACIXOYTUXAW-UHFFFAOYSA-N
Synonym 2-bromoacetophenone, phenacyl bromide, 2-bromo-1-phenylethan-1-one, bromoacetophenone, ethanone, 2-bromo-1-phenyl, bromomethyl phenyl ketone, alpha-bromoacetophenone, stauffer 4644, omega-bromoacetophenone, acetophenone, 2-bromo
PubChem CID 6259
ChEBI CHEBI:51846
IUPAC Name 2-bromo-1-phenylethanone
SMILES C1=CC=C(C=C1)C(=O)CBr
View More Specs

Products 4
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10357420
View Documents
Thermo Scientific Acros
152010250
25 g Plastic bottle
32.86 €
25g
Please to purchase this item. Need a web account? Register with us today!
10071720
View Documents
Thermo Scientific Acros
152011000
100 g Plastic bottle
70.75 €
100g
Please to purchase this item. Need a web account? Register with us today!
10635012
View Documents
Thermo Scientific Acros
152015000
500 g Plastic bottle
237.18 €
500g
Please to purchase this item. Need a web account? Register with us today!
10669853
View Documents
Thermo Scientific Acros
152010025
2.5 kg Plastic bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Acylation reagent
Specifications

Chemical Identifiers

70-11-1
199.05
LIGACIXOYTUXAW-UHFFFAOYSA-N
6259
2-bromo-1-phenylethanone
C8H7BrO
MFCD00000195
2-bromoacetophenone, phenacyl bromide, 2-bromo-1-phenylethan-1-one, bromoacetophenone, ethanone, 2-bromo-1-phenyl, bromomethyl phenyl ketone, alpha-bromoacetophenone, stauffer 4644, omega-bromoacetophenone, acetophenone, 2-bromo
CHEBI:51846
C1=CC=C(C=C1)C(=O)CBr

Specifications

70-11-1
100.0
Brown-Green to White
>112°C
97.5% min. (GC)
C6H5COCH2Br
07, 283
15, 1409
Solubility in water: practically insoluble in water. Other solubilities: freely soluble in alcohol,benzene,chloroform,and ether
C1=CC=C(C=C1)C(=O)CBr
199.05
CHEBI:51846
98%
2-Bromoacetophenone, 98%
97.5
46.0°C to 51.0°C
135.0°C (18.0 mmHg)
Authentic
C8H7BrO
MFCD00000195
02,316; 12,381
2-bromoacetophenone, phenacyl bromide, 2-bromo-1-phenylethan-1-one, bromoacetophenone, ethanone, 2-bromo-1-phenyl, bromomethyl phenyl ketone, alpha-bromoacetophenone, stauffer 4644, omega-bromoacetophenone, acetophenone, 2-bromo
LIGACIXOYTUXAW-UHFFFAOYSA-N
2-bromo-1-phenylethanone
6259
199.05
Crystals or Powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Toxic if inhaled.
Causes severe skin burns and eye damage.
Toxic if swallowed.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 200-724-9

missing translation for 'tsca' : TSCA

Product Identifier
  • 2-Bromoacetophenone

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H331-Toxic if inhaled.
  • H301-Toxic if swallowed.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.