missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-5-nitrobenzonitrile, 97%, Thermo Scientific™

26.20 € - 82.50 €

Chemical Identifiers

CAS 134604-07-2
Molecular Formula C7H3BrN2O2
Molecular Weight (g/mol) 227.017
MDL Number MFCD00234249
InChI Key RKODNVITKISFKU-UHFFFAOYSA-N
Synonym 1-cyano-2-bromo-5-nitrobenzene, 2-bromo-1-cyano-5-nitrobenzene, benzonitrile, 2-bromo-5-nitro, 2-bromo-5-nitro-benzonitrile, acmc-209bwc, bromo-5-nitrobenzonitrile, 2-bromo-5-nitrobenzo nitrile, ksc494g7d, 2-bromo-5-ni tro-benzonitrile, 2-bromo-5-nitrobenzenecarbonitrile
PubChem CID 7095423
IUPAC Name 2-bromo-5-nitrobenzonitrile
SMILES C1=CC(=C(C=C1[N+](=O)[O-])C#N)Br
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15431988
View Documents Promotion Details
Thermo Scientific Alfa Aesar
H55671.03
1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
15441988
View Documents Promotion Details
Thermo Scientific Alfa Aesar
H55671.06
5 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

134604-07-2
227.017
RKODNVITKISFKU-UHFFFAOYSA-N
7095423
C1=CC(=C(C=C1[N+](=O)[O-])C#N)Br
C7H3BrN2O2
MFCD00234249
1-cyano-2-bromo-5-nitrobenzene, 2-bromo-1-cyano-5-nitrobenzene, benzonitrile, 2-bromo-5-nitro, 2-bromo-5-nitro-benzonitrile, acmc-209bwc, bromo-5-nitrobenzonitrile, 2-bromo-5-nitrobenzo nitrile, ksc494g7d, 2-bromo-5-ni tro-benzonitrile, 2-bromo-5-nitrobenzenecarbonitrile
2-bromo-5-nitrobenzonitrile

Specifications

134604-07-2
C7H3BrN2O2
Moisture sensitive
RKODNVITKISFKU-UHFFFAOYSA-N
2-bromo-5-nitrobenzonitrile
7095423
97%
118°C to 120°C
MFCD00234249
1-cyano-2-bromo-5-nitrobenzene, 2-bromo-1-cyano-5-nitrobenzene, benzonitrile, 2-bromo-5-nitro, 2-bromo-5-nitro-benzonitrile, acmc-209bwc, bromo-5-nitrobenzonitrile, 2-bromo-5-nitrobenzo nitrile, ksc494g7d, 2-bromo-5-ni tro-benzonitrile, 2-bromo-5-nitrobenzenecarbonitrile
C1=CC(=C(C=C1[N+](=O)[O-])C#N)Br
227.017
227.02
2-Bromo-5-nitrobenzonitrile
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

missing translation for 'tsca' : No

Recommended Storage : Keep cold

SDS
Documents

Documents

Product Certifications