missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4'-methylacetophenone, 97%, Thermo Scientific Chemicals

Product Code. 10464735
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Plastic bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 619-41-0
Molecular Formula C9H9BrO
Molecular Weight (g/mol) 213.07
MDL Number MFCD00000203
InChI Key KRVGXFREOJHJAX-UHFFFAOYSA-N
Synonym 2-bromo-4'-methylacetophenone, 2-bromo-1-p-tolyl ethanone, 4-methylphenacyl bromide, 2-bromo-1-4-methylphenyl ethanone, 2-bromo-1-p-tolylethanone, 2-bromo-4-methylacetophenone, 2-bromo-1-4-methylphenyl ethan-1-one, acetophenone, 2-bromo-4'-methyl, ethanone, 2-bromo-1-4-methylphenyl, p-methylphenacyl bromide
PubChem CID 69272
IUPAC Name 2-bromo-1-(4-methylphenyl)ethanone
SMILES CC1=CC=C(C=C1)C(=O)CBr

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 52°C to 53°C
Color White to Yellow
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Plastic bottle
Linear Formula CH3C6H4COCH2Br
Quantity 100 g
Beilstein 07, 309
Formula Weight 213.07
Percent Purity 97%
Physical Form Crystalline Powder or Needles
Chemical Name or Material 2-Bromo-4'-methylacetophenone
Show More Show Less
compliance-icons
Product Identifier
  • 2-Bromo-4'-methylacetophenone
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.