missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4'-chloroacetophenone, 98+%, Thermo Scientific Chemicals

50.10 € - 138.00 €

Chemical Identifiers

CAS 536-38-9
Molecular Formula C8H6BrClO
Molecular Weight (g/mol) 233.49
MDL Number MFCD00000625
InChI Key FLAYZKKEOIAALB-UHFFFAOYSA-N
Synonym 2-bromo-4'-chloroacetophenone, 2-bromo-1-4-chlorophenyl ethanone, 4-chlorophenacyl bromide, p-chlorophenacyl bromide, 2-bromo-1-4-chlorophenyl ethan-1-one, ethanone, 2-bromo-1-4-chlorophenyl, 4'-chlorophenacyl bromide, 2-bromo-p-chloroacetophenone, p-bromoacetyl chlorobenzene, 4'-chloro-2-bromoacetophenone
PubChem CID 68303
IUPAC Name 2-bromo-1-(4-chlorophenyl)ethanone
SMILES C1=CC(=CC=C1C(=O)CBr)Cl
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10667492
View Documents Promotion Details
Thermo Scientific Acros
146360250
25 g Glass bottle
50.10 €
25g
Please to purchase this item. Need a web account? Register with us today!
10021660
View Documents Promotion Details
Thermo Scientific Acros
146361000
100 g Glass bottle
138.00 €
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

536-38-9
233.49
FLAYZKKEOIAALB-UHFFFAOYSA-N
68303
C1=CC(=CC=C1C(=O)CBr)Cl
C8H6BrClO
MFCD00000625
2-bromo-4'-chloroacetophenone, 2-bromo-1-4-chlorophenyl ethanone, 4-chlorophenacyl bromide, p-chlorophenacyl bromide, 2-bromo-1-4-chlorophenyl ethan-1-one, ethanone, 2-bromo-1-4-chlorophenyl, 4'-chlorophenacyl bromide, 2-bromo-p-chloroacetophenone, p-bromoacetyl chlorobenzene, 4'-chloro-2-bromoacetophenone
2-bromo-1-(4-chlorophenyl)ethanone

Specifications

536-38-9
100.0
White to Yellow
Authentic
C8H6BrClO
MFCD00000625
15, 2156
FLAYZKKEOIAALB-UHFFFAOYSA-N
2-bromo-1-(4-chlorophenyl)ethanone
68303
98+%
2-Bromo-4′-chloroacetophenone, 98+%
98.0
95.0°C to 99.0°C
1% max. (70°C, 2 hr.)
98% min. (GC)
ClC6H4COCH2Br
07, 285
2-bromo-4'-chloroacetophenone, 2-bromo-1-4-chlorophenyl ethanone, 4-chlorophenacyl bromide, p-chlorophenacyl bromide, 2-bromo-1-4-chlorophenyl ethan-1-one, ethanone, 2-bromo-1-4-chlorophenyl, 4'-chlorophenacyl bromide, 2-bromo-p-chloroacetophenone, p-bromoacetyl chlorobenzene, 4'-chloro-2-bromoacetophenone
C1=CC(=CC=C1C(=O)CBr)Cl
233.49
233.49
Crystals
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 208-631-5

missing translation for 'rtecsNumber' : AM5978800

missing translation for 'tsca' : TSCA

Product Identifier
  • 2-Bromo-4'-chloroacetophenone

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only