missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-3'-hydroxyacetophenone, 96%, Thermo Scientific Chemicals

88.90 € - 302.00 €

Chemical Identifiers

CAS 2491-37-4
Molecular Formula C8H7BrO2
Molecular Weight (g/mol) 215.04
MDL Number MFCD05664326
InChI Key IEPSGFQQGKPTPM-UHFFFAOYSA-N
PubChem CID 6917206
IUPAC Name 2-bromo-1-(3-hydroxyphenyl)ethanone
SMILES C1=CC(=CC(=C1)O)C(=O)CBr
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10169969
View Documents
Thermo Scientific Acros
428620010
1 g Glass bottle
88.90 €
1g
Please to purchase this item. Need a web account? Register with us today!
10301670
View Documents
Thermo Scientific Acros
428620050
5 g Glass bottle
302.00 €
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

2491-37-4
215.04
IEPSGFQQGKPTPM-UHFFFAOYSA-N
2-bromo-1-(3-hydroxyphenyl)ethanone
C8H7BrO2
MFCD05664326
6917206
C1=CC(=CC(=C1)O)C(=O)CBr

Specifications

2491-37-4
100.0
95% min. (HPLC)
(HO)C6H4COCH2Br
2-bromo-1-3-hydroxyphenyl ethanone, 2-bromo-3'-hydroxyacetophenone, 2-bromo-3-hydroxyacetophenone, ethanone, 2-bromo-1-3-hydroxyphenyl, 3-hydroxyphenacyl bromide, 2-bromo-1-3-hydroxyphenyl ethan-1-one, ethanone,2-bromo-1-3-hydroxyphenyl, 2-bromo-3`-hydroxyacetophenone, 2-bromo-3'-hydroxy-acetophenone, tos-bb-1139
C1=CC(=CC(=C1)O)C(=O)CBr
215.04
215.04
2-Bromo-3′-hydroxyacetophenone
95.0
70°C to 74°C
C8H7BrO2
MFCD05664326
IEPSGFQQGKPTPM-UHFFFAOYSA-N
2-bromo-1-(3-hydroxyphenyl)ethanone
6917206
96%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

Product Identifier
  • 2-Bromo-3'-hydroxyacetophenone

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.