missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-1-(4-chloro-3-nitrophenyl)ethan-1-one, Technical Grade, Thermo Scientific™

95.00 € - 484.00 €

Chemical Identifiers

CAS 22019-49-4
Molecular Formula C8H5BrClNO3
Molecular Weight (g/mol) 278.486
InChI Key FXNQTMRPLLNPQY-UHFFFAOYSA-N
PubChem CID 2778330
IUPAC Name 2-bromo-1-(4-chloro-3-nitrophenyl)ethanone
SMILES C1=CC(=C(C=C1C(=O)CBr)[N+](=O)[O-])Cl
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10427622
View Documents Promotion Details
Thermo Scientific Maybridge
JP00700DA
1 g Amber Glass Bottle
95.00 €
1g
Please to purchase this item. Need a web account? Register with us today!
10334192
View Documents Promotion Details
Thermo Scientific Maybridge
JP00700EA
10 g Amber Glass Bottle
243.00 €
10g
Please to purchase this item. Need a web account? Register with us today!
10327014
View Documents Promotion Details
Thermo Scientific Maybridge
JP00700EB
25 g Amber Glass Bottle
484.00 €
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

22019-49-4
278.486
2778330
C1=CC(=C(C=C1C(=O)CBr)[N+](=O)[O-])Cl
C8H5BrClNO3
FXNQTMRPLLNPQY-UHFFFAOYSA-N
2-bromo-1-(4-chloro-3-nitrophenyl)ethanone

Specifications

22019-49-4
C8H5BrClNO3
FXNQTMRPLLNPQY-UHFFFAOYSA-N
2-bromo-1-(4-chloro-3-nitrophenyl)ethanone
2778330
95%
95%
2-bromo-1-4-chloro-3-nitrophenyl ethanone, 2-bromo-1-4-chloro-3-nitrophenyl ethan-1-one, 4-chloro-3-nitrophenacylbromide, 4-chloro-3-nitrophenacyl bromide, ethanone,2-bromo-1-4-chloro-3-nitrophenyl, 4-chloro-3-nitro-phenacyl bromide, 4-chloro-3-nitrophen-acyl bromide, 2-bromo-4'-chloro-3'-nitroacetophenone, 2-bromo-1-4-chloro-3-nitro-phenyl ethanone, ethanone, 2-bromo-1-4-chloro-3-nitrophenyl
C1=CC(=C(C=C1C(=O)CBr)[N+](=O)[O-])Cl
278.486
278.49
2-Bromo-1-(4-chloro-3-nitrophenyl)ethan-1-one
Videos
SDS
Documents

Documents

Product Certifications