missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-1-(3-phenylisoxazol-5-yl)ethan-1-one 95%, Thermo Scientific™

49.00 € - 222.00 €

Chemical Identifiers

CAS 14731-14-7
Molecular Formula C11H8BrNO2
Molecular Weight (g/mol) 266.09
MDL Number MFCD00173898
InChI Key XTBXGZOVSCTNEC-UHFFFAOYSA-N
Synonym 5-bromoacetyl-3-phenylisoxazole, 2-bromo-1-3-phenylisoxazol-5-yl ethan-1-one, 2-bromo-1-3-phenylisoxazol-5-yl ethanone, 2-bromo-1-3-phenyl-1,2-oxazol-5-yl ethanone, 2-bromo-1-3-phenyl-1,2-oxazol-5-yl ethan-1-one, 2-bromo-1-3-phenyl-isoxazol-5-yl-ethanone, ethanone, 2-bromo-1-3-phenyl-5-isoxazolyl, pubchem8717, buttpark 43\57-72
PubChem CID 2735532
IUPAC Name 2-bromo-1-(3-phenyl-1,2-oxazol-5-yl)ethanone
SMILES BrCC(=O)C1=CC(=NO1)C1=CC=CC=C1
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10089481
View Documents
Thermo Scientific Maybridge
SPB04811DA
1 g Amber Glass Bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10062264
View Documents
Thermo Scientific Maybridge
SPB04811EA
10 g Amber Glass Bottle
222.00 €
10g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

14731-14-7
266.09
XTBXGZOVSCTNEC-UHFFFAOYSA-N
2735532
BrCC(=O)C1=CC(=NO1)C1=CC=CC=C1
C11H8BrNO2
MFCD00173898
5-bromoacetyl-3-phenylisoxazole, 2-bromo-1-3-phenylisoxazol-5-yl ethan-1-one, 2-bromo-1-3-phenylisoxazol-5-yl ethanone, 2-bromo-1-3-phenyl-1,2-oxazol-5-yl ethanone, 2-bromo-1-3-phenyl-1,2-oxazol-5-yl ethan-1-one, 2-bromo-1-3-phenyl-isoxazol-5-yl-ethanone, ethanone, 2-bromo-1-3-phenyl-5-isoxazolyl, pubchem8717, buttpark 43\57-72
2-bromo-1-(3-phenyl-1,2-oxazol-5-yl)ethanone

Specifications

14731-14-7
C11H8BrNO2
5-bromoacetyl-3-phenylisoxazole, 2-bromo-1-3-phenylisoxazol-5-yl ethan-1-one, 2-bromo-1-3-phenylisoxazol-5-yl ethanone, 2-bromo-1-3-phenyl-1,2-oxazol-5-yl ethanone, 2-bromo-1-3-phenyl-1,2-oxazol-5-yl ethan-1-one, 2-bromo-1-3-phenyl-isoxazol-5-yl-ethanone, ethanone, 2-bromo-1-3-phenyl-5-isoxazolyl, pubchem8717, buttpark 43\57-72
BrCC(=O)C1=CC(=NO1)C1=CC=CC=C1
266.09
266.09
2-Bromo-1-(3-phenylisoxazol-5-yl)ethan-1-one
95%
MFCD00173898
XTBXGZOVSCTNEC-UHFFFAOYSA-N
2-bromo-1-(3-phenyl-1,2-oxazol-5-yl)ethanone
2735532
95%
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.