missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromo-1-(2,4-dimethylphenyl)ethan-1-one, Thermo Scientific™
40.30 € - 159.60 €
Chemical Identifiers
CAS | 26346-85-0 |
---|---|
Molecular Formula | C10H11BrO |
Molecular Weight (g/mol) | 227.101 |
InChI Key | GSCCVWPVPFIRJP-UHFFFAOYSA-N |
Synonym | 2-bromo-1-2,4-dimethylphenyl ethanone, 2-bromo-1-2,4-dimethylphenyl ethan-1-one, ethanone, 2-bromo-1-2,4-dimethylphenyl, 2-bromo-2',4'-dimethylacetophenone, 2,4-dimethylphenacyl bromide, 2,4-dimethyl phenacyl bromide, pubchem16796, 2,4-dimethylphenacyl bromid, ksc557o1l, alpha-bromo-2',4'-dimethylacetophenone |
PubChem CID | 2063450 |
IUPAC Name | 2-bromo-1-(2,4-dimethylphenyl)ethanone |
SMILES | CC1=CC(=C(C=C1)C(=O)CBr)C |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10130692
|
Thermo Scientific Maybridge
CD00248DA |
1 g | Amber Glass Bottle |
40.30 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10293022
|
Thermo Scientific Maybridge
CD00248EA |
10 g | Amber Glass Bottle | N/A | |||||
10422074
|
Thermo Scientific Maybridge
CD00248EB |
25 g | Amber Glass Bottle | N/A | |||||
Description
Chemical Identifiers
26346-85-0 | |
227.101 | |
2-bromo-1-2,4-dimethylphenyl ethanone, 2-bromo-1-2,4-dimethylphenyl ethan-1-one, ethanone, 2-bromo-1-2,4-dimethylphenyl, 2-bromo-2',4'-dimethylacetophenone, 2,4-dimethylphenacyl bromide, 2,4-dimethyl phenacyl bromide, pubchem16796, 2,4-dimethylphenacyl bromid, ksc557o1l, alpha-bromo-2',4'-dimethylacetophenone | |
2-bromo-1-(2,4-dimethylphenyl)ethanone |
C10H11BrO | |
GSCCVWPVPFIRJP-UHFFFAOYSA-N | |
2063450 | |
CC1=CC(=C(C=C1)C(=O)CBr)C |
Specifications
26346-85-0 | |
C10H11BrO | |
2-bromo-1-2,4-dimethylphenyl ethanone, 2-bromo-1-2,4-dimethylphenyl ethan-1-one, ethanone, 2-bromo-1-2,4-dimethylphenyl, 2-bromo-2',4'-dimethylacetophenone, 2,4-dimethylphenacyl bromide, 2,4-dimethyl phenacyl bromide, pubchem16796, 2,4-dimethylphenacyl bromid, ksc557o1l, alpha-bromo-2',4'-dimethylacetophenone | |
CC1=CC(=C(C=C1)C(=O)CBr)C | |
227.101 | |
227.1 | |
2-Bromo-1-(2, 4-dimethylphenyl)ethan-1-one |
97% | |
(CH3)2C6H3C(O)CH2Br | |
GSCCVWPVPFIRJP-UHFFFAOYSA-N | |
2-bromo-1-(2,4-dimethylphenyl)ethanone | |
2063450 | |
97% |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title