missing translation for 'onlineSavingsMsg'
Learn More

2-Benzyloxybenzeneboronic acid, 96%, Thermo Scientific Chemicals

48.50 € - 159.00 €

Chemical Identifiers

CAS 190661-29-1
Molecular Formula C13H13BO3
Molecular Weight (g/mol) 228.054
MDL Number MFCD01632206
InChI Key MCAIDINWZOCYQK-UHFFFAOYSA-N
Synonym 2-benzyloxyphenylboronic acid, 2-benzyloxy phenylboronic acid, 2-benzyloxybenzeneboronic acid, 2-benzyloxy phenyl boronic acid, 2-benzyloxyphenyl boronic acid, 2-benzyloxyphenylborornic acid, 2-benzyloxy phenyl boranediol, o-benzyloxyphenylboronic acid, boronic acid, 2-phenylmethoxy phenyl
PubChem CID 2773253
IUPAC Name (2-phenylmethoxyphenyl)boronic acid
SMILES B(C1=CC=CC=C1OCC2=CC=CC=C2)(O)O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11374536
View Documents Promotion Details
Thermo Scientific Alfa Aesar
L20100.03
1 g
48.50 €
1g
Please to purchase this item. Need a web account? Register with us today!
11384536
View Documents Promotion Details
Thermo Scientific Alfa Aesar
L20100.06
5 g
159.00 €
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

190661-29-1
228.054
MCAIDINWZOCYQK-UHFFFAOYSA-N
2773253
B(C1=CC=CC=C1OCC2=CC=CC=C2)(O)O
C13H13BO3
MFCD01632206
2-benzyloxyphenylboronic acid, 2-benzyloxy phenylboronic acid, 2-benzyloxybenzeneboronic acid, 2-benzyloxy phenyl boronic acid, 2-benzyloxyphenyl boronic acid, 2-benzyloxyphenylborornic acid, 2-benzyloxy phenyl boranediol, o-benzyloxyphenylboronic acid, boronic acid, 2-phenylmethoxy phenyl
(2-phenylmethoxyphenyl)boronic acid

Specifications

190661-29-1
C13H13BO3
7813704
MCAIDINWZOCYQK-UHFFFAOYSA-N
(2-phenylmethoxyphenyl)boronic acid
2773253
96%
96°C to 99°C
MFCD01632206
2-benzyloxyphenylboronic acid, 2-benzyloxy phenylboronic acid, 2-benzyloxybenzeneboronic acid, 2-benzyloxy phenyl boronic acid, 2-benzyloxyphenyl boronic acid, 2-benzyloxyphenylborornic acid, 2-benzyloxy phenyl boranediol, o-benzyloxyphenylboronic acid, boronic acid, 2-phenylmethoxy phenyl
B(C1=CC=CC=C1OCC2=CC=CC=C2)(O)O
228.054
228.06
2-Benzyloxybenzeneboronic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only