missing translation for 'onlineSavingsMsg'
Learn More

2-(Aminomethyl)indole, 97%, Thermo Scientific Chemicals

66.03 € - 985.00 €

Chemical Identifiers

CAS 21109-25-1
Molecular Formula C9H10N2
Molecular Weight (g/mol) 146.193
MDL Number MFCD03422512
InChI Key RNAODKZCUVVPEN-UHFFFAOYSA-N
Synonym 1h-indol-2-ylmethyl amine, 1h-indol-2-yl methanamine, 1h-indole-2-methanamine, 1-1h-indol-2-yl methanamine, 2-aminomethyl indole, 2-aminomethylindole, c-1h-indol-2-yl-methylamine, 2-indolemethylamine, 1h-indol-2-yl methylamine, indol-2-ylmethylamine
PubChem CID 582331
IUPAC Name 1H-indol-2-ylmethanamine
SMILES C1=CC=C2C(=C1)C=C(N2)CN
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15490598
SDS View Documents
Thermo Scientific Alfa Aesar
H54281.MD
250 mg
66.03 €
250mg
Please to purchase this item. Need a web account? Register with us today!
15470598
SDS View Documents
Thermo Scientific Alfa Aesar
H54281.03
1 g
209.04 €
1g
Please to purchase this item. Need a web account? Register with us today!
15480598
SDS View Documents
Thermo Scientific Alfa Aesar
H54281.06
5 g N/A N/A N/A
Description

Description

2-(Aminomethyl)indole is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(Aminomethyl)indole is used as a pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
Specifications

Chemical Identifiers

21109-25-1
146.193
RNAODKZCUVVPEN-UHFFFAOYSA-N
582331
C1=CC=C2C(=C1)C=C(N2)CN
C9H10N2
MFCD03422512
1h-indol-2-ylmethyl amine, 1h-indol-2-yl methanamine, 1h-indole-2-methanamine, 1-1h-indol-2-yl methanamine, 2-aminomethyl indole, 2-aminomethylindole, c-1h-indol-2-yl-methylamine, 2-indolemethylamine, 1h-indol-2-yl methylamine, indol-2-ylmethylamine
1H-indol-2-ylmethanamine

Specifications

21109-25-1
MFCD03422512
Slightly soluble in water.
C1=CC=C2C(=C1)C=C(N2)CN
146.193
146.19
2-(Aminomethyl)indole
C9H10N2
1h-indol-2-ylmethyl amine, 1h-indol-2-yl methanamine, 1h-indole-2-methanamine, 1-1h-indol-2-yl methanamine, 2-aminomethyl indole, 2-aminomethylindole, c-1h-indol-2-yl-methylamine, 2-indolemethylamine, 1h-indol-2-yl methylamine, indol-2-ylmethylamine
RNAODKZCUVVPEN-UHFFFAOYSA-N
1H-indol-2-ylmethanamine
582331
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-(Aminomethyl)indole, 97%, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.