Learn More
2'-Aminoacetophenone, 98%, Thermo Scientific Chemicals
38.09 € - 710.54 €
Chemical Identifiers
CAS | 551-93-9 |
---|---|
Molecular Formula | C8H9NO |
Molecular Weight (g/mol) | 135.17 |
MDL Number | MFCD00007717 |
InChI Key | GTDQGKWDWVUKTI-UHFFFAOYSA-N |
Synonym | 2'-aminoacetophenone, 1-2-aminophenyl ethanone, o-aminoacetophenone, 2-acetylaniline, o-acetylaniline, o-aminoacetylbenzene, 1-acetyl-2-aminobenzene, 1-2-aminophenyl ethan-1-one, acetophenone, 2'-amino, ethanone, 1-2-aminophenyl |
PubChem CID | 11086 |
IUPAC Name | 1-(2-aminophenyl)ethanone |
SMILES | CC(=O)C1=CC=CC=C1N |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11309689
|
Thermo Scientific Alfa Aesar
A10895.06 |
5 g |
38.09 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11464253
|
Thermo Scientific Alfa Aesar
A10895.18 |
50 g |
177.89 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11474253
|
Thermo Scientific Alfa Aesar
A10895.30 |
250 g |
710.54 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
551-93-9 | |
135.17 | |
GTDQGKWDWVUKTI-UHFFFAOYSA-N | |
11086 | |
CC(=O)C1=CC=CC=C1N |
C8H9NO | |
MFCD00007717 | |
2'-aminoacetophenone, 1-2-aminophenyl ethanone, o-aminoacetophenone, 2-acetylaniline, o-acetylaniline, o-aminoacetylbenzene, 1-acetyl-2-aminobenzene, 1-2-aminophenyl ethan-1-one, acetophenone, 2'-amino, ethanone, 1-2-aminophenyl | |
1-(2-aminophenyl)ethanone |
Specifications
551-93-9 | |
253°C to 255°C | |
C8H9NO | |
MFCD00007717 | |
14,413 | |
GTDQGKWDWVUKTI-UHFFFAOYSA-N | |
1-(2-aminophenyl)ethanone | |
11086 | |
98% |
1.118 | |
112°C (233°F) | |
1.616 | |
386122 | |
2'-aminoacetophenone, 1-2-aminophenyl ethanone, o-aminoacetophenone, 2-acetylaniline, o-acetylaniline, o-aminoacetylbenzene, 1-acetyl-2-aminobenzene, 1-2-aminophenyl ethan-1-one, acetophenone, 2'-amino, ethanone, 1-2-aminophenyl | |
CC(=O)C1=CC=CC=C1N | |
135.17 | |
135.17 | |
2'-Aminoacetophenone |
Safety and Handling
GHS H Statement
H302-H315-H319
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
missing translation for 'einecsNumber' : 209-002-8
missing translation for 'tsca' : Yes
Recommended Storage : Keep cold
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.